SAB5702410
Anti-Purinergic Receptor P2Y6 Antibody, clone 4I8C9, Rabbit Monoclonal
Synonym(s):
P2Y6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
Recommended Products
biological source
rabbit
Quality Level
material
colorless
clone
4I8C9, monoclonal
form
liquid
mol wt
42 kDa
species reactivity
human
concentration
0.58 mg/mL
technique(s)
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
VAQERRGKAARMAVVVAAAFAISFLPFHITKTAYLAVRSTPGVPCTVLEAFAAAYKGTRPFASANSVLDPILFYFTQKKFRRRPHELLQKLTAKWQRQGR
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... P2RY6(5031)
General description
The product of this gene belongs to the family of P2 receptors, which is activated by extracellular nucleotides and subdivided into P2X ligand-gated ion channels and P2Y G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor, which is a G-protein coupled receptor, is responsive to UDP, partially responsive to UTP and ADP, and not responsive to ATP. It is proposed that this receptor mediates inflammatory responses. Alternative splicing results in multiple transcript variants that encode different protein isoforms. [provided by RefSeq, Mar 2013]
Immunogen
A synthetic peptide corresponding to a sequence within amino acids 229-328 of human Purinergic Receptor P2Y6 (Q15077).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Application
WB
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service