Skip to Content
Merck
All Photos(2)

Documents

SAB4501670

Sigma-Aldrich

Anti-Keratin 19 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CK-19, Cytokeratin-19, K19, KRT19, Keratin type I cytoskeletal 19

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 44 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT19(3880)

General description

Anti-Keratin 19 Antibody detects endogenous levels of total Keratin 19 protein.
Cytokeratin 19 (KRT19) is a cytoplasmic intermediate filament protein with a α-helical central domain. It belongs to the acidic type I cytokeratin family of proteins (KRTs). This gene is mapped to human chromosome 17q21.

Immunogen

The antiserum was produced against synthesized peptide derived from human Keratin 19.

Immunogen Range: 231-280

Biochem/physiol Actions

Cytokeratin 19 (KRT19) plays a major role in structural rigidity and multipurpose scaffolds. Overexpression of this gene in various cancers may participate in tumorigenic transformation. It may modulate signaling pathways and mediate cell apoptosis, cell cycle arrest, invasion and metastasis. KRT19 plays a vital role in the cancer stemness of hepatocellular carcinoma (HCC).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytokeratin 19 (KRT19) has a role in the reprogramming of cancer stem cell-like cells to less aggressive and more drug-sensitive cells
Saha S, et al.
International Journal of Molecular Sciences, 19(5), 1423-1423 (2018)
The tissue-dependent keratin 19 gene transcription is regulated by GKLF/KLF4 and Sp1
Brembeck FH and Rustgi AK
Test, 275(36), 28230-28239 (2000)
Cansu Tekin et al.
Molecular oncology, 15(11), 3091-3108 (2021-05-02)
Pancreatic acinar cells have high plasticity and can transdifferentiate into ductal-like cells. This acinar-to-ductal metaplasia (ADM) contributes to tissue maintenance but may also contribute to the premalignant transformation that can eventually progress to pancreatic ductal adenocarcinoma (PDAC). Macrophages are key

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service