Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500869

Sigma-Aldrich

Anti-CD32 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD32, CDW32, FC γ receptor IIB, FC-γ RII, FC-γ RII-B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 34 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FCGR2B(2213)

General description

The human immune system has six FcγRs-FcγRI, FcγRIIA, FcγRIIB, FcγRIIC, FcγRIIIA and FcγRIIIB. FcγRIIIB/CD32 (Fc fragment of IgG receptor IIb) is an inhibitory receptor that is located on human chromosome 1q23.

Anti-CD32 Antibody detects endogenous levels of total CD32 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CD32.

Immunogen Range: 277-326

Biochem/physiol Actions

FcγRIIIB/CD32 (Fc fragment of IgG receptor IIb) controls the generation of autoantibodies and their effector roles. During a viral memory response, FcγRIIB blocks the generation of inflammatory type I IFN from plasmacytoid dendritic cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impairment on the lateral mobility induced by structural changes underlies the functional deficiency of the lupus-associated polymorphism Fc?RIIB-T232
Xu L, et al.
The Journal of Experimental Medicine (2016)
Fc?RIIB and autoimmunity
Espeli M, et al.
Immunological Reviews (2016)
Fc?RIIB prevents inflammatory type I IFN production from plasmacytoid dendritic cells during a viral memory response
Flores M, et al.
Journal of Immunology (2015)
Association of a transmembrane polymorphism of Fcgamma receptor IIb (FCGR2B) with systemic lupus erythematosus in Taiwanese patients
Chen JY, et al.
Arthritis and Rheumatism (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service