Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500487

Sigma-Aldrich

Anti-PARP (Cleaved-Asp214) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ADP-ribosyltransferase 1, ADPRT, ADPRT1, PARP, pADPRT-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 24 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Asp214)

Gene Information

human ... PARP1(142)

General description

Anti-PARP (Cleaved-Asp214) Antibody detects endogenous levels of fragment of activated PARP (Cleaved-Asp214) protein.
PARP1 (poly (ADP-ribose) polymerase 1) is a nuclear protein that consists of three functional domains. It belongs to the PARP family. It is located on chromosome 1q42.

Immunogen

The antiserum was produced against synthesized peptide derived from human PARP.

Immunogen Range: 165-214

Application

Anti-PARP (Cleaved-Asp214) antibody has been used in Western Blotting.

Biochem/physiol Actions

PARP1 (poly (ADP-ribose) polymerase 1) participates in DNA repair. It serves as a facilitator of homologous recombination. It controls transcription by regulating chromatin structure, altering DNA methylation patterns, acting as a co-regulator of transcription factors and interacting with chromatin insulators. It also plays an important role in the protection of cardiovascular system.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PARP-1 protein expression in glioblastoma multiforme
Galia A, et al.
European Journal of Histochemistry, 56 (2012)
PARP inhibition: PARP1 and beyond
Rouleau M, et al.
Nature Reviews. Cancer, 10 (2010)
Trapping of PARP1 and PARP2 by Clinical PARP Inhibitors
Murai J, et al.
Cancer Research, 72 (2012)
Fascin2 regulates cisplatin-induced apoptosis in NRK-52E cells
Wang X, et al.
Toxicology Letters (2017)
Loss of ?(E)-catenin potentiates cisplatin-induced nephrotoxicity via increasing apoptosis in renal tubular epithelial cells
Wang X, et al.
Toxicological Sciences, 141 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service