Skip to Content
Merck
All Photos(2)

Documents

SAB4300033

Sigma-Aldrich

Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-APRF antibody produced in rabbit, Anti-FLJ20882 antibody produced in rabbit, Anti-HIES antibody produced in rabbit, Anti-MGC16063 antibody produced in rabbit, Anti-signal transducer and activator of transcription 3 (acute-phase response factor) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~88 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(A-P-YP-L-K)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr705)

Gene Information

human ... STAT3(6774)

Related Categories

General description

Signal transducer and activator of transcription 3 (STAT3) is a member of STAT family and it composed of 770 amino acids. The gene encoding protein consists of 24 exons and is extended over a length of 75170 bases on genomic DNA STAT3 is mapped on human chromosome 17q21.2. The protein is characterized with a conserved amino-terminal domain, a DNA-binding domain, an SH2 domain and a carboxy-terminal transactivation domain.

Immunogen

Peptide sequence around phosphorylation site of tyrosine 705 (A-P-Y(p)-L-K), according to the protein STAT3.

Application

Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit has been used in immunohistochemical staining and immunoblotting.

Biochem/physiol Actions

Signal transducer and activator of transcription 3 (STAT3) plays a vital role in various cellular responses such as induction of an acute-phase response in hepatoma cells, stimulation of proliferation in B lymphocytes, activation of terminal differentiation and growth arrest in monocytes, and regulation of the pluripotency of embryonic stem cells. Activated STAT3 has been involved in malignant transformation. STAT3 may be a key player in the pathogenesis of diverse human cancers which makes this molecule a prime target for novel therapies. Phospho-tyrosine residue 705 (Tyr705) Stat3 can be used as a potential marker for determining overall survival of node-negative breast cancer patients. Phosphorylated STAT3 might be involved in angiogenesis, anti-apoptosis and tumor progression.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Transcription factor that binds to the interleukin-6 (IL-6)-responsive elements identified in the promoters of various acute-phase protein genes. Activated by IL31 through IL31RA.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phospho-Stat3 expression and correlation with VEGF, p53, and Bcl-2 in gastric carcinoma using tissue microarray.
Choi JH
Acta Pathologica, Microbiologica et Immunological Scandinavica, 114, 619-625 (2006)
Porphyromonas gingivalis and its lipopolysaccharide differently modulate epidermal growth factor-dependent signaling in human gingival epithelial cells.
Elkaim R
Journal of Oral Microbiology, 9 (2017)
Liyun Chen et al.
The Journal of clinical investigation, 133(15) (2023-06-06)
Patients with cancer who have high serum levels of squamous cell carcinoma antigen 1 (SCCA1, now referred to as SERPINB3) commonly experience treatment resistance and have a poor prognosis. Despite being a clinical biomarker, the modulation of SERPINB3 in tumor
R Elkaim et al.
Journal of oral microbiology, 9(1), 1334503-1334503 (2017-07-28)
Periodontitis is an inflammatory disease induced by pathogenic bacteria such as
Manchao Zhang et al.
Molecular cancer therapeutics, 14(10), 2313-2320 (2015-08-01)
Epithelial-restricted with serine box (ESX), a member of the ETS transcription factor family, is elevated and regulates EGFR in head and neck squamous cell carcinoma (HNSCC). However, the molecular mechanisms that contribute to ESX dysregulation remain to be elucidated. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service