Skip to Content
Merck
All Photos(2)

Key Documents

SAB2500639

Sigma-Aldrich

Anti-MOG antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-MGC26137, Anti-Myelin oligodendrocyte glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

bovine, mouse, rat, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MOG(4340)

General description

Myelin/oligodendrocyte glycoprotein (MOG) is an integral membrane glycoprotein belonging to the large immunoglobulin superfamily. It is a 26-28kDa glycoprotein localized on chromosome 6p21.3. It consists of an extracellular immunoglobulin-like domain and two hydrophobic regions. It is expressed on the surface of myelinating oligodendrocytes and external lamellae of myelin sheaths in the central nervous system.

Immunogen

Peptide with sequence C-AGQFLEELRNPF from the C Terminus of the protein sequence according to NP_996532.2; NP_996535.2; NP_001008229.1.

Application

Anti-MOG antibody produced in goat is suitable for indirect ELISA and western blot applications.

Biochem/physiol Actions

Myelin/oligodendrocyte glycoprotein (MOG) is a major component of the central nervous system. It is involved in several cellular processes including cellular adhesion, in the regulation of oligodendrocyte microtubule stability and facilitates the interactions between myelin and the immune system. It has been reported that MOG may be involved in the completion and/or maintenance of the myelin sheath, and in cell-cell communication. It has also been observed that mutation or polymorphisms in MOG gene could cause multiple sclerosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B della Gaspera et al.
European journal of biochemistry, 258(2), 478-484 (1999-01-05)
Myelin/oligodendrocyte glycoprotein (MOG), a specific component of the mammalian central nervous system, is located on the surface of the oligodendrocyte plasma membrane and the outermost lamellae of mature myelin; it is expressed during the latter steps of myelinogenesis. It has
D Pham-Dinh et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(17), 7990-7994 (1993-09-01)
Myelin/oligodendrocyte glycoprotein (MOG) is found on the surface of myelinating oligodendrocytes and external lamellae of myelin sheaths in the central nervous system, and it is a target antigen in experimental autoimmune encephalomyelitis and multiple sclerosis. We have isolated bovine, mouse
D Pham-Dinh et al.
Genomics, 29(2), 345-352 (1995-09-20)
Myelin/oligodendrocyte glycoprotein (MOG), a specific component of the central nervous system localized on the outermost lamellae of mature myelin, is a member of the immunoglobulin superfamily. We report here the organization of the human MOG gene, which spans approximately 17
T G Johns et al.
Journal of neurochemistry, 72(1), 1-9 (1999-01-14)
Myelin oligodendrocyte glycoprotein (MOG) is a quantitatively minor component of CNS myelin whose function remains relatively unknown. As MOG is an autoantigen capable of producing a demyelinating multiple sclerosis-like disease in mice and rats, much of the research directed toward

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service