Skip to Content
Merck
All Photos(1)

Documents

SAB1303190

Sigma-Aldrich

ANTI-SLC8A1 (CENTER) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

CNC, NCX1, Na(+)/Ca(2+)-exchange protein 1, SLC8A1, Sodium/calcium exchanger 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

108547 Da

species reactivity

human, mouse

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:50-1:100
western blot: 1:250-1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC8A1(6546)

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongxin Li et al.
International urology and nephrology, 51(6), 1059-1070 (2019-04-24)
Previous studies indicate that autosomal dominant polycystic kidney disease (ADPKD) cells exhibited dysregulated calcium homeostasis and enhanced cell proliferation. TRPC3 has been shown to function in the modulation of calcium and sodium entry, but whether TRPC3 plays a role in
Shigeki Moriguchi et al.
Molecular neurobiology, 56(6), 4381-4394 (2018-10-17)
Na+/Ca2+ exchangers (NCXs) are mainly expressed in the plasma membrane and exchange one Ca2+ for three Na+, depending on the electrochemical gradients across the plasma membrane. NCXs have three isoforms, NCX1-3, encoded by distinct genes in mammals. Here, we report
Xing Huang et al.
Autophagy, 15(7), 1258-1279 (2019-02-23)
Notwithstanding the numerous drugs available for liver cancer, emerging evidence suggests that chemotherapeutic resistance is a significant issue. HGF and its receptor MET play critical roles in liver carcinogenesis and metastasis, mainly dependent on the activity of receptor tyrosine kinase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service