Skip to Content
Merck
All Photos(3)

Key Documents

S9007

Sigma-Aldrich

Spectinomycin dihydrochloride pentahydrate

potency: ≥603 μg per mg

Synonym(s):

Actinospectacin, Actinospectacin dihydrochloride pentahydrate, Spectinomycin, Spectinomycin HCL, Spectinomycin Hydrochloride, Spectinomycin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24N2O7 · 2HCl · 5H2O
CAS Number:
Molecular Weight:
495.35
EC Number:
MDL number:
UNSPSC Code:
51281666
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

synthetic (inorganic)

Quality Level

form

powder

potency

≥603 μg per mg

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white to pale buff

solubility

H2O: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

agriculture

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

O.O.O.O.O.Cl.Cl.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

InChI

1S/C14H24N2O7.2ClH.5H2O/c1-5-4-6(17)14(20)13(21-5)22-12-10(19)7(15-2)9(18)8(16-3)11(12)23-14;;;;;;;/h5,7-13,15-16,18-20H,4H2,1-3H3;2*1H;5*1H2/t5-,7-,8+,9+,10+,11-,12-,13+,14+;;;;;;;/m1......./s1

InChI key

DCHJOVNPPSBWHK-UXXUFHFZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Spectinomycin is an aminocyclitol antibiotic derived from Streptomyces spectabilis. It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr, to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons. It is recommended for use in cell culture applications at 7.5-20 mg/L.

Biochem/physiol Actions

Mode of Action: Spectinomycin acts by inhibiting protein synthesis and elongation by binding to the bacterial 30S ribosomal subunit and interfering with peptidyl tRNA translocation.

Mode of Resistance: A mutation in rpsE, the gene for ribosomal protein S5, prevents the binding of spectinomycin. Resistance is also conferred by the aminoglycoside-3′-adenyltransferase gene (aadA).

Antimicrobial spectrum: Spectinomycin acts against gram-negative and gram-positive bacteria.

Caution

Solutions can be stored at 2-8°C for several weeks or at -20°C for more extended periods.

Preparation Note

Stock solutions should be prepared at 10 mg/mL in water and filter sterilized.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amber M Hotto et al.
G3 (Bethesda, Md.), 1(7), 559-570 (2012-03-03)
Noncoding RNAs (ncRNA) are widely expressed in both prokaryotes and eukaryotes. Eukaryotic ncRNAs are commonly micro- and small-interfering RNAs (18-25 nt) involved in posttranscriptional gene silencing, whereas prokaryotic ncRNAs vary in size and are involved in various aspects of gene
Stefan Schwarz et al.
The Journal of antimicrobial chemotherapy, 53(2), 379-382 (2004-01-20)
Prior to the renewal of spectinomycin licensing for veterinary uses in Germany, 154 Pasteurella multocida and 148 Mannheimia haemolytica strains from respiratory tract infections in cattle were investigated for their MICs of spectinomycin and other antimicrobial agents. The data obtained
Yi-Ywan M Chen et al.
Applied and environmental microbiology, 77(12), 3967-3974 (2011-05-03)
Streptococcus parasanguinis is among the most successful colonizers of the human body. Strain FW213 harbors a 7.0-kb cryptic plasmid, pFW213, with a copy number at 5 to 10 per chromosome. Sequence and functional analyses of pFW213 revealed that the open
Janice Yee Chi Lo et al.
The Journal of antimicrobial chemotherapy, 67(6), 1422-1426 (2012-02-16)
To undertake laboratory and clinical surveillance of gonococcal antimicrobial susceptibility to various therapeutic agents in Hong Kong, so as to monitor for emerging resistance and to inform on appropriate choice of empirical therapy. Trends in susceptibility of gonococci to ceftriaxone
Magnus Unemo et al.
Antimicrobial agents and chemotherapy, 57(2), 1057-1061 (2012-11-28)
Gonorrhea may become untreatable, and new treatment options are essential. Verified resistance to spectinomycin is exceedingly rare. However, we describe a high-level spectinomycin-resistant (MIC, >1,024 μg/ml) Neisseria gonorrhoeae strain from Norway with a novel resistance mechanism. The resistance determinant was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service