Skip to Content
Merck
All Photos(2)

Key Documents

RAB0366

Sigma-Aldrich

Mouse MMP-2 ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 25 pg/mL
standard curve range: 27.43-20000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Mmp2(17390)

General description

Matrix Metallopeptidase 2 (MMP-2), also known as gelatinase A, belongs to the gelatinase subclass of the matrix metalloproteinase (MMP) family. It contains zinc in the active site. MMP-2 cleaves gelatin, type IV, V, VII, X, and XI collagens, fibronectin, vitronectin, elastin, laminin, proteoglycans, and a range of non-extracellular matrix (ECM) components. Both MMP-2 and MMP9 are important for matrix degradation and they are implicated in the maintenance of neovascularization. The Mouse MMP-2 enzyme-linked immunosorbent assay (ELISA) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse MMP-2 in serum, plasma, cell culture supernatants, and urine.

Immunogen

Recombinant Mouse MMP2

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Mouse MMP-2 ELISA Kit has been used in enzyme-linked immunosorbent assay to quantify matrix metalloproteinase -2 (MMP-2) secreted in supernatants by gold nanoparticles (AuNP)-loaded endothelial cells and superparamagnetic iron oxide nanoparticle (SPION) loaded endothelial cells.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhadeep Chakrabarti et al.
Experimental lung research, 31(6), 599-621 (2005-07-16)
The lung is affected by a variety of disease processes that can lead to considerable morbidity and mortality. As the lung is the only organ for respiration and gas exchange, the structural and functional integrity of the lung is of
Takayuki Shiomi et al.
Pathology international, 60(7), 477-496 (2010-07-03)
Cellular functions within tissues are strictly regulated by the tissue microenvironment which comprises extracellular matrix and extracellular matrix-deposited factors such as growth factors, cytokines and chemokines. These molecules are metabolized by matrix metalloproteinases (MMP), a disintegrin and metalloproteinases (ADAM) and
J C Rodriguez-Manzaneque et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(22), 12485-12490 (2001-10-19)
Growth of tumors and metastasis are processes known to require neovascularization. To ascertain the participation of the endogenous angiogenic inhibitor thrombospondin-1 (TSP1) in tumor progression, we generated mammary tumor-prone mice that either lack, or specifically overexpress, TSP1 in the mammary
Disturbance of adhesomes by gold nanoparticles reveals a size- and cell type-bias.
Mulens-Arias, et al.
Biomaterials Science, 7, 389-408 (2019)
Theranostic iron oxide nanoparticle cargo defines extracellular vesicle-dependent modulation of macrophage activation and migratory behavior
Mulens-Arias, V, et al.
Advanced Biosystems, 1800079-1800079 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service