Skip to Content
Merck
All Photos(2)

Key Documents

RAB0283

Sigma-Aldrich

Human IL-1ra ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 100 pg/mL
standard curve range: 0.124-30 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL1RN(3557)

General description

The Human IL-1ra ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-1ra in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL-1 Receptor Antagonist

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Interleukin-1 receptor antagonist (IL-1RA) inhibits the activity of IL-1α and IL-1β by competitively binding to their type I and type II receptors. IL-1RA plays a vital role in inhibition of acute pro-inflammatory response and forbidding chronic inflammation from damaging healthy tissues. Abnormal expression of IL-1RA leads to development of inflammatory or autoimmune related diseases. Allelic polymorphism of IL-1RA leads to inflammatory bowel disease (IBD). Therapeutically, IL-1RA may help in the treatment of sepsis, rheumatoid arthritis, asthma. Overexpression of IL-1RA is observed in serum of psoriatic arthritis (PsA) patients.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum levels of IL-10, IL-6, IL-1ra, and sIL-2R in patients with psoriatic arthritis.
Elkayam O
Rheumatology International, 19(3), 101-105 (2000)
Expression of interleukin-1 beta (IL-1beta) and interleukin-1 receptor antagonist (IL-1ra) on asthmatic bronchial epithelium.
Sousa AR
American Journal of Respiratory and Critical Care Medicine, 154(4 Pt 1), 1061-1066 (1996)
A novel association between IL1-Ra (receptor antagonist) gene polymorphism and T1DM in Al-Madina Al-Mounawra.
Ali RA
European Review for Medical and Pharmacological Sciences, 19(19), 3701-3708 (2015)
Allelic polymorphism in IL-1 beta and IL-1 receptor antagonist (IL-1Ra) genes in inflammatory bowel disease.
Bioque G
Clinical and Experimental Immunology, 102(2), 379-383 (1995)
Relationship of plasma leptin to plasma cytokines and human survivalin sepsis and septic shock.
Arnalich F
The Journal of Infectious Diseases, 180(3), 908-911 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service