Skip to Content
Merck
All Photos(1)

Key Documents

MTOXCE2C9

Sigma-Aldrich

CypExpress 2C9 Cytochrome P450 human

Synonym(s):

CPC9, CYP2C, CYP2C10, CYP2C9, CYPIIC9, P450 2C9, P450IIC9

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41106200
NACRES:
NA.77

biological source

human

recombinant

expressed in Pichia pastoris

form

dry powder

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP2C9(1559)

Biochem/physiol Actions

Cytochrome P450 enzymes are heme containing monooxygenases which are found primarily in the mammalian liver. They catalyze the oxidative metabolism of xenobiotics. This metabolism is the initial step in the biotransformation and elimination of a wide variety of drugs and environmental pollutants from the body. Sulfaphenazole, fluconazole, fluvoxamine and fluoxetine are inhibitors, whereas rifampin is an inducer of the enzyme. The isozyme CYP2C9 is a major pathway for the invivo conversion of Diclofenac to 4-Hydroxydiclofenac via hydroxylation.
Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Some isoforms have narrow substrate specificity, while others are promiscuous.

Features and Benefits

CypExpress 2C9 is a permeabilized and stabilized dried yeast powder preparation containing full length, unmodified, human CYP 2C9 and recombinant human NADPH oxidoreductase. Using proprietary processes, CypExpress are expressed in Pichia yeast. Following optimal expression, the cells are inactivated, permeabilized and dried to a fine powder. CypExpress retains the cellular mechanisms to provide the P450 enzymes with the energy and cofactors to continue to function for long experiments, and can generate larger amounts of metabolite than mammalian microsomes or other genetically engineered expression systems. It is designed for rapid screening of drug metabolites and for scaling up metabolite generation.

Unit Definition

A reaction mixture containing 500 μM diclofenac and 100 mg/mL of CypExpress 2C9 will produce 428 μM 4-hydroxydiclofenac in 2 hours at 30°C under the conditions described.

Physical form

Light yellow, very fine powder

Reconstitution

Per Protocol/Variable

Legal Information

CypExpress is a trademark of Oxford Biomedical Research

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service