MTCB12005
MTC™ Bio Western Blot Box
L × W × D 2 7/8 in. (7.3 cm) × 2 in. (5.1 cm) × 1 1/4 in. (3.2 cm), clear
Synonym(s):
multi blot box, stacking western blot box
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
Recommended Products
material
clear
polystyrene
packaging
pack of 5 ea
manufacturer/tradename
MTC™ Bio B1200-5
L × W × D
2 7/8 in. (7.3 cm) × 2 in. (5.1 cm) × 1 1/4 in. (3.2 cm)
Looking for similar products? Visit Product Comparison Guide
General description
Blot Box, 2 7/8 x 2 x 1 1/4in. (7.3 x 5.1 x 3.2cm), 5/pk
These Blot boxes are correctly matched to common blot sizes, enabling more blots to fit on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. All corners are formed at 90˚ angles and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Black Blot boxes are light-blocking opaque for use with light sensitive samples.
Application
Half Mini-Protein Gels
Features and Benefits
- A perfect box for every blot
- Reduce antibody usage
- Hinged lid minimizes evaporation
- For halves, wholes, strips or minis
Legal Information
MTC is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service