MAK447
Alkaline Phosphatase Assay Kit
Sufficient for 250 colorimetric tests
Synonym(s):
ALP Activity Assay Kit, Alkaline Phosphatase Test Kit
About This Item
Recommended Products
Quality Level
application(s)
pharmaceutical
detection method
colorimetric
relevant disease(s)
osteoporosis; gastrointestinal diseases; orthopedic diseases; cancer
storage temp.
−20°C
General description
Application
- Cancer Research
- Gastroenterology Disease Research
- Osteoporosis Research
Features and Benefits
Simplified Process: Experience a streamlined process with the addition of only a single working reagent and a short 5 minute assay reaction, reducing complexity and saving valuable time and effort.
Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.
Suitability
Principle
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2
Target Organs
Nervous system,Kidney,Liver,Skin
Storage Class Code
8A - Combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service