Skip to Content
Merck
All Photos(5)

Key Documents

HPA011055

Sigma-Aldrich

Anti-SNN antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AG8_1, Anti-Stannin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

EDEESIVGDGETKEPFLLVQYSAKGPCVERKAKLMTPNGPEV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNN(8303)

General description

SNN (stannin) is a mitochondrial membrane protein, which has a membrane helical region, a linker region containing conserved CXC metal-binding motif, a predictive 14-3-3ζ binding domain and a cytosolic helical region. This gene is localized to human chromosome 16p13, and has an open readin frame of 264bp. The mRNA of SNN is present in hippocampus, neocortex, cerebellum, striatum, midbrain, lung, spleen and kidney, and is absent in heart, liver, skeletal muscle and testis. This protein is made of 88 amino acids and has a molecular weight of 10kDa.

Immunogen

Stannin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SNN (stannin) is involved in the toxicity of organotin compounds in cells, such as trimethyltin (TMT), though it is not sufficient alone to mediate toxicity. This protein is also responsible for neuronal cell death mediated by TMT toxicity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72342

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N S Dejneka et al.
Neurochemistry international, 31(6), 801-815 (1997-12-31)
The cDNA encoding the protein stannin was isolated previously via subtractive hybridization, using differential expression after trimethyltin (TMT) intoxication, as a basis for isolating mRNA which may be expressed in TMT-sensitive cells. Initial characterization revealed a novel gene product which
S M Toggas et al.
Molecular pharmacology, 42(1), 44-56 (1992-07-01)
The molecular basis of selective vulnerability of specific neuronal populations to neurotoxicants remains a key focus in neurotoxicology. Trimethyltin (TMT) selectively damages neurons in rodent and human central nervous system after a single exposure. By coupling subtractive hybridization with molecular
N S Dejneka et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 9(7), 556-564 (1998-07-10)
Stannin is a protein that has been localized to trimethyltin-sensitive cell populations, and evidence suggests it plays a role in the toxic effects of organotins. In this study, we have isolated a mouse stannin genomic clone and have characterized the
Bethany A Buck-Koehntop et al.
Journal of molecular biology, 354(3), 652-665 (2005-10-26)
Organotin compounds or alkyltins are ubiquitous environmental toxins that have been implicated in cellular death. Unlike other xenobiotic compounds, such as organomercurials and organoleads, alkyltins activate apoptotic cascades at low concentrations. Trimethyltin (TMT) chloride is amongst the most toxic organotin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service