Skip to Content
Merck
All Photos(4)

Key Documents

HPA002725

Sigma-Aldrich

Anti-DMD antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

VVCQQSATQRLEHAFNIARYQLGIEKLLDPEDVDTTYPDKKSILMYITSLFQVLPQQVSIEAIQEVEMLPRPPKVTKEEHFQLHHQMHYSQQITVSLAQGYERTSSPKPRFKSYAYTQAAYVTTSDPTRSPFPSQHLEAPEDKSFGSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DMD(1756)

General description

Dystrophin (DMD) is a submembranous cytoskeletal protein. It plays a major role in various processes of skeletal, cardiac and smooth muscles as well as of peripheral and central nervous systems, including the retina.

Immunogen

Dystrophin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In striated muscle cells, DMD (dystrophin) acts as a component of the multimeric protein dystrophin-associated glycoprotein (DGC) complex. It helps DGC to adhere with the extracellular matrix for various signalling pathways. It helps to maintain structural assembly of plasma membrane, regulates ion homeostasis and transmembrane signaling cascade. It stabilzes muscle fiber cells. It is associated with various muscular dystrophies, such as Duchenne/Becker muscular dystrophy, congenital muscular dystrophy and different forms of limb-girdle muscular dystrophy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74322

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ueda et al.
Histology and histopathology, 15(3), 753-760 (2000-08-30)
Dramatical development of molecular genetics has been disclosing the molecular mechanism of Duchenne muscular dystrophy (DMD) and Becker muscular dystrophy (BMD). DMD gene product, dystrophin, is a submembranous cytoskeletal protein and many dystrophin-associated proteins (DAPs) have been identified, such as
K G Culligan et al.
International journal of molecular medicine, 2(6), 639-648 (1998-12-16)
The membrane cytoskeletal component dystrophin and its associated glycoproteins play a central role in the molecular pathogenesis of several muscular dystrophies, i.e. Duchenne/Becker muscular dystrophy, congenital muscular dystrophy and various forms of limb-girdle muscular dystrophy. Although the most frequent of
Downregulation of Dystrophin Expression Occurs across Diverse Tumors, Correlates with the Age of Onset, Staging and Reduced Survival of Patients.
Alnassar, et al.
Cancers, 15 (2023)
T Haenggi et al.
Cellular and molecular life sciences : CMLS, 63(14), 1614-1631 (2006-05-20)
The dystrophin glycoprotein complex (DGC) is a multimeric protein assembly associated with either the X-linked cytoskeletal protein dystrophin or its autosomal homologue utrophin. In striated muscle cells, the DGC links the extracellular matrix to the actin cytoskeleton and mediates three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service