Skip to Content
Merck
All Photos(2)

Key Documents

H8416

Sigma-Aldrich

Stem Cell Factor human

≥95% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

Synonym(s):

SCF, c-Kit ligand

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Stem Cell Factor human, SCF, recombinant, expressed in HEK 293 cells, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤200 ng/mL ED50

quality

endotoxin tested

mol wt

dimer 35-45 kDa (glycosylated)

packaging

pkg of 5x10 μg
pkg of 10 μg
pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... KITLG(4254)

Looking for similar products? Visit Product Comparison Guide

General description

The KITLG (KIT ligand) gene is mapped to human chromosome 12q21.32.

Biochem/physiol Actions

Stem Cell Factor (SCF), also known as c-Kit ligand (KL), steel factor (SLF) and mast cell growth factor (MGF), is a 30 kDa glycoprotein with broad activities on various tissues, including hematopoietic cells, pigment cells, and primordial germ cells.
Stem Cell Factor (SCF), also known as c-Kit ligand (KL), steel factor (SLF) and mast cell growth factor (MGF), is a 30 kDa glycoprotein with broad activities on various tissues, including hematopoietic cells, pigment cells, and primordial germ cells. SCF is secreted by endothelial cells, fibroblasts, and bone marrow stromal cells as a membrane-bound form which may be cleaved to release the soluble form. Both forms are active in promoting colony formation from murine bone marrow cells, but membrane-bound SCF is more effective in promoting hematopoieses in vivo, suggesting a role in cellular interactions between hematopoietic and stromal cells. The soluble is thought to exist in solution as a noncovalently linked dimer. SCF is structurally related to M-CSF (CSF-1) and Flt-3/Flk-2 Ligand (FL) with all three sharing a similar size, existence of transmembrane and soluble forms, four conserved cysteines, and alternative splicing exon locations, but they share little sequence homology. SCF alone is a modest colony stimulating factor. However, in the presence of other cytokines such as EPO, TPO, GM-CSF, G-CSF, M-CSF, IL-3, and IL-7, SCF is a potent costimulant that works synergistically to increase the size of myeloid, erythroid or lymphoid lineage colonies without influencing the lineage differentiation of the progenitors.
Stem cell factor (SCF) of granulosa cell origin, efficiently contributes to the follicular development and oocyte maturation. SCF controls tumor development and progression. Elevated SCF gene expression is observed in hepatocellular carcinoma.

Preparation Note

SCF is expressed in human HEK 293 cells as a glycosylated monomer with an apparent molecular mass of 35-45 kDa due to glycosylation. Production in human 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The specific activity was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells (human erythroleukemic indicator cell line).

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene mapping study for constitutive skin color in an isolated Mongolian population
Paik SH, et al.
Experimental & Molecular Medicine, 44, 241-249 (2012)
Increased SCF in Follicular Fluid and Granulosa Cells Positively Correlates With Oocyte Maturation, Fertilization, and Embryo Quality in Humans.
Tan J, et al.
Reproductive Sciences, 24(11), 1544-1550 (2017)
Shaoshao Zhang et al.
Frontiers in pharmacology, 13, 932092-932092 (2022-08-26)
Aims: Calcific aortic valve disease (CAVD) is a chronic cardiovascular disease with high morbidity that lacks effective pharmacotherapeutics. As a natural flavonoid extracted from Ampelopsis grossedentata, dihydromyricetin (DHM) has been shown to be effective in protecting against atherosclerosis; yet, the
Yoshimichi Okayama et al.
Immunologic research, 34(2), 97-115 (2006-06-09)
Mast cells play a pivotal role in immediate hypersensitivity and chronic allergic reactions that can contribute to asthma, atopic dermatitis, and other allergic diseases. Because mast cell numbers are increased at sites of inflammation in allergic diseases, pharmacologic intervention into
Prabir Ray et al.
Cell cycle (Georgetown, Tex.), 7(18), 2826-2832 (2008-09-13)
The receptor tyrosine kinase, c-kit, and its ligand, stem cell factor (SCF), function in a diverse range of biological functions. The role of c-kit in the maintenance and survival of hematopoietic stem cells and of mast cells is well recognized.

Articles

Read article on hematopoietic cytokines and hematopoiesis

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service