GE17-0010-01
Sephadex® G-10
Cytiva 17-0010-01, pack of 100 g
Synonym(s):
Sephadex G-10 column, Sephadex gel
About This Item
Recommended Products
feature
autoclavable In wet form (pH 7.0) at 120°C for 30 min
packaging
pack of 100 g
manufacturer/tradename
Cytiva 17-0010-01
parameter
flow rate (The beads behave as rigid spheres obeying Darcy′s Law.)
matrix
cross-linked dextran
particle size
40-120 μm (dry)
55-165 (wet)
working range
2-13
General description
Application
Features and Benefits
- Quickly desalts, removes contaminants and transfers to a new buffer in a single step.
- Suitable for buffer exchange and cleanup of biological samples, for example peptides, small Proteins, and oligosaccharides larger than 700 molecular weight
- Efficient removal of contaminants such as salts, dyes, and radioactive labels
- Lowest exclusion limit (700 molecular weight) in the Sephadex® G-range
Storage and Stability
Analysis Note
Legal Information
Storage Class Code
13 - Non Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
This page shows how to perform a separation with a Sephadex column from Cytiva which are Size Exclusion Chromatography media consisting of an epichlorohydrin cross-linked dextran.
Column Packing and Preparation for Size Exclusion Chromatography
Related Content
Sephadex® from Cytiva and Darcy’s law in Size Exclusion Chromatography
Separation options for Size Exclusion Chromatography with Sephadex from Cytiva.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service