Skip to Content
Merck
All Photos(1)

Key Documents

EHU061641

Sigma-Aldrich

MISSION® esiRNA

targeting human UVRAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTGAGCACCTCAAACTTCAACTCCAGAAGGAATCCCTAAATGAGCTGAGGAAGGAGTGCACTGCAAAAAGAGAACTCTTCTTGAAGACTAATGCTCAGTTGACAATTCGTTGCAGGCAGTTACTCTCTGAGCTTTCCTACATTTACCCTATTGATTTGAATGAACATAAGGATTACTTTGTATGCGGTGTCAAGTTGCCTAATTCTGAGGACTTCCAAGCAAAAGATGATGGAAGCATTGCTGTTGCCCTTGGTTATACTGCACATCTGGTCTCCATGATTTCCTTTTTCCTACAAGTGCCCCTCAGATATCCTATAATTCATAAGGGGTCTAGATCAACAATCAAAGACAATATCAATGACAAACTGACGGAAAAGGAGAGAGAGTTTCCACTGTATCCAAAAGGAGGGGAGAAGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoyuan Deng et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 41(6), 2171-2182 (2017-04-26)
Atherosclerosis is a multifactorial chronic disease and is the main cause of death and impairment in the world. Endothelial injury and apoptosis play a crucial role in the onset and development of atherosclerosis. MicroRNAs (miRNAs) have been proven to be
Jianwen Wang et al.
OncoTargets and therapy, 13, 10275-10285 (2020-10-30)
Radiotherapy is one of the most important methods in the treatment of patients with hypopharyngeal squamous cell carcinoma (HSCC). However, radioresistance will be developed after repeated irradiation. Among many key factors contributing to radioresistance, enhanced autophagy is recognized as one
Yunha Kim et al.
Autophagy, 11(5), 796-811 (2015-05-07)
The EWSR1 (EWS RNA-binding protein 1/Ewing Sarcoma Break Point Region 1) gene encodes a RNA/DNA binding protein that is ubiquitously expressed and involved in various cellular processes. EWSR1 deficiency leads to impairment of development and accelerated senescence but the mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service