Skip to Content
Merck
All Photos(1)

Documents

D4196

Sigma-Aldrich

Dehydroleucodine

≥98% (HPLC)

Synonym(s):

11,13-Dehydroleucodin, Dehydroleucodin, Leucodin, Lidbeckialactone, Mesatlantin E, NSC 180034, [3aS-(3aalpha,9aalpha,9bbeta)]-3,3a,4,5,9a,9b-hexahydro-6,9-dimethyl-3-methylene-azuleno[4,5-b]furan-2,7-dione, dehydro-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O3
CAS Number:
Molecular Weight:
244.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to off-white

solubility

DMSO: ≥30 mg/mL

storage temp.

room temp

SMILES string

CC1=C2[C@@H]([C@H]3OC(=O)C(=C)[C@@H]3CC1)C(C)=CC2=O

InChI

1S/C15H16O3/c1-7-4-5-10-9(3)15(17)18-14(10)13-8(2)6-11(16)12(7)13/h6,10,13-14H,3-5H2,1-2H3/t10-,13-,14-/m0/s1

InChI key

SKNVIAFTENCNGB-BPNCWPANSA-N

Biochem/physiol Actions

Dehydroleucodine is a sesquiterpene lactone isolated from Artemisia douglasiana Besser; mast cell stabilizers. An extract from the plant is widely used in Cuyo region (Argentina) as folk medicine for the treatment of gastric ailments. The compound prevents gastric and duodenal damages in response to necrosis-inducing agents. Stabilization of mast cells may be a key mechanism to protect the gastrointestinal tract from injury and ulceration. Recently, it was established that dehydroleucodine and xanthatin inhibit mast cell degranulation induced by compound 48/80.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariano E Vera et al.
Leukemia & lymphoma, 53(9), 1795-1803 (2012-02-04)
The aim of the present study was to determine whether dehydroleucodine, xanthatin and 3-benzyloxymethyl-5H-furan-2-one inhibit the activation of human leukemic LAD2 mast cells induced by compound 48/80 or the calcium ionophore A23187. LAD2 cells were preincubated in the presence of
G H Wendel et al.
Fitoterapia, 79(1), 1-5 (2007-08-09)
Dehydroleucodine (DhL), a sesquiterpene lactone obtained from Artemisia douglasiana, was screened for antidiarrheal effects. DhL inhibited castor oil-induced diarrhea in mice by judged by a decrease in the number of wet faeces in the DhL-treatment groups. DhL significantly reduced intestinal
G Sánchez Toranzo et al.
Zygote (Cambridge, England), 15(2), 183-187 (2007-04-28)
In amphibian oocytes meiosis, the transition from G2 to M phase is regulated by the maturation promoting factor (MPF), a complex of the cyclin-dependent kinase p34/cdc2 and cyclin B. In immature oocytes there is an inactive complex (pre-MPF), in which
Adriana Galvis et al.
European journal of pharmacology, 671(1-3), 18-25 (2011-10-04)
Dehydroleucodine (DhL) is a sesquiterpene lactone of the guaianolide group with gastric cytoprotective activity. Recent studies have also demonstrated that DhL inhibits the proliferation of vascular smooth muscle cells. In this study we examined the effect of DhL in the
Alicia Penissi et al.
Cells, tissues, organs, 173(4), 234-241 (2003-05-27)
Dehydroleucodine (DhL), a sesquiterpene lactone isolated from Artemisia douglasiana Besser, prevents gastroduodenal damage elicited by necrosis-inducing agents such as absolute ethanol. Changes in the number of mast cells or evidence of activation of the cells for mediator release have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service