Skip to Content
Merck

Skip To

C7017

Choline chloride

≥99%

Synonym(s):

(2-Hydroxyethyl)trimethylammonium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)3N(Cl)CH2CH2OH
CAS Number:
Molecular Weight:
139.62
UNSPSC Code:
50501514
NACRES:
NA.25
PubChem Substance ID:
EC Number:
200-655-4
Beilstein/REAXYS Number:
3563126
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

≥99%

form

powder

color

white

mp

302-305 °C (dec.) (lit.)

lipid type

phospholipids

SMILES string

[Cl-].C[N+](C)(C)CCO

InChI

1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

SGMZJAMFUVOLNK-UHFFFAOYSA-M

General description

Choline acetyltransferase substrate
Choline is a constituent of sphingomyelin and lecithin. It is a precursor of acetylcholine. Choline plays a vital role in methyl group metabolism, carcinogenesis and lipid transport. Choline deficiency is associated with fatty liver. It maintains cell structural integrity and cell signalling. Choline is implicated in the synthesis of phospholipids. It acts as a potent biomarker for ischemic heart disease.

Application

Choline chloride has been used:
  • in choline release assay
  • as an endogenous agonist of sigma-1 receptors (Sig-1Rs)
  • as a standard to analyse interrelationships between methionine and choline metabolism

Biochem/physiol Actions

Choline is an essential nutrient, commonly grouped with the B complex vitamins, that plays key roles in many biological processes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2697826978PHG0021
biological source

synthetic

biological source

synthetic

biological source

synthetic

biological source

synthetic

assay

≥99%

assay

≥99.0% (AT)

assay

≥99.0% (AT)

assay

-

form

powder

form

powder or crystals

form

powder or crystals

form

powder

mp

302-305 °C (dec.) (lit.)

mp

302-305 °C (dec.) (lit.)

mp

302-305 °C (dec.) (lit.)

mp

302-305 °C (dec.) (lit.)

color

white

color

colorless to white

color

colorless to white

color

white

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

400

biological source

synthetic

biological source

synthetic

biological source

synthetic

biological source

synthetic

assay

≥99%

assay

≥99.0% (AT)

assay

≥99.0% (AT)

assay

-

form

powder

form

powder or crystals

form

powder or crystals

form

powder

mp

302-305 °C (dec.) (lit.)

mp

302-305 °C (dec.) (lit.)

mp

302-305 °C (dec.) (lit.)

mp

302-305 °C (dec.) (lit.)

color

white

color

colorless to white

color

colorless to white

color

white

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

400


Still not finding the right product?

Explore all of our products under Choline chloride


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
C7017-25G04061833511787
C7017-10MG04061833511770
C7017-100G04061833511763
C7017-5G04061833511794

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters