Key Documents
BMSIPS2050PT4
Sigma-Aldrich
Benchmark Overhead Stirrer Propeller
4-arm, PTFE-coated
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
material
PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-P-T4
diam. × L
65 mm × 300 mm
General description
4-arm PTFE coated propeller is useful for mid to high speed mixing.
Stiring rods and propellers to fit Benchmark range of OS-20L and OS-40L overhead stirrers.
Application
stirring, mixing