Skip to Content
Merck
All Photos(1)

Key Documents

B9433

Sigma-Aldrich

Bovine Serum, Adult

USA origin, sterile-filtered, suitable for cell culture

Synonym(s):

Bovine sera, cow sera, cow serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160409
NACRES:
NA.71
biological source:
bovine
origin:
USA origin
sterility:
sterile-filtered
form:
liquid
shipped in:
dry ice

biological source

bovine

Quality Level

sterility

sterile-filtered

form

liquid

composition

hemoglobin, ≤50 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤50 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

General description

Bovine serum (BS) is considered a cost-effective choice than fetal bovine serum (FBS). It can also be preserved stable for a long time.

Application

Bovine Serum, Adult has been used:
  • to treat silicone square to prepare it for the biofilm assay for in vitro biofilm growth conditions
  • in cell culture as a component in Dulbecco′s modified Eagle medium for digesting articular cartilage and proximal metacarpals
  • as a lubricating solution in hydrodynamic analysis and to imitate the synovial joints

Biochem/physiol Actions

Adult bovine serum (BS) possesses bactericidal activity against Helicobacter pylori. Heat-treated BS can be used as the best serum source of H. pylori culture medium.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Pakkanen
In vitro cellular & developmental biology. Animal, 30A(5), 295-299 (1994-05-01)
A mixture containing an ultrafiltrate fraction (UF) of bovine colostrum (6.7%), adult bovine serum (BS) (1%), and human holo-transferrin (hTF) (5 mg/liter) was developed for cultivation of Chinese hamster ovary cells (CHO-K1) and African green monkey kidney cells (Vero). The
Steven H Elder et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 24(4), 740-747 (2006-03-04)
Cells are often cultured at high density (e.g., confluent monolayer and as pellets) to promote chondrogenic differentiation and to maintain the chondrocyte phenotype. They are also frequently suspended in hydrogels such as agarose or alginate for the same purposes. These
Study on surface texture patterns for improving tribological performance of bioimplants
Shen G, et al.
Surface and Coatings Technology (2021)
Priya Uppuluri et al.
Mycopathologia, 168(3), 101-109 (2009-04-17)
Candida albicans biofilms on most medical devices are exposed to a flow of body fluids that provide water and nutrients to the fungal cells. While C. albicans biofilms grown in vitro under static conditions have been exhaustively studied, the same
D Nečas et al.
Journal of the mechanical behavior of biomedical materials, 69, 66-74 (2017-01-04)
The aim of the present study is to provide an analysis of protein film formation in hip joint replacements considering real conformity based on in situ observation of the contact zone. The main attention is focused on the effect of

Articles

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service