Skip to Content
Merck
All Photos(2)

Key Documents

AV35165

Sigma-Aldrich

Anti-KCND3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Potassium voltage-gated channel, Shal-related subfamily, member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

rabbit, rat, guinea pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCND3(3752)

General description

KCND3 is a voltage-gated potassium (Kv) channel that is involved in the repolarization of the action potential. Genetic alterations in KCND3 have been associated with Brugada syndrome and sudden unexplained death.
Rabbit Anti-KCND3 antibody recognizes human, mouse, rat, rabbit, and pig KCND3.

Immunogen

Synthetic peptide directed towards the middle region of human KCND3

Application

Rabbit Anti-KCND3 antibody is suitable for western blot applications at a concentration of 1.25 μg/ml.

Biochem/physiol Actions

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). KCND3 encodes a member of the potassium channel, voltage-gated, shal-related subfamily, members of which form voltage-activated A-type potassium ion channels and are prominent in the repolarization phase of the action potential.

Sequence

Synthetic peptide located within the following region: KARLARIRVAKTGSSNAYLHSKRNGLLNEALELTGTPEEEHMGKTTSLIE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John R Giudicessi et al.
Human mutation, 33(6), 989-997 (2012-03-30)
Heritable arrhythmia syndromes, including Brugada syndrome (BrS) and idiopathic ventricular fibrillation (IVF), may serve as the pathogenic basis for autopsy-negative sudden unexplained death (SUD) and sudden infant death syndrome (SIDS). Emerging evidence has linked perturbations in the transient outward current
John R Giudicessi et al.
Heart rhythm, 8(7), 1024-1032 (2011-02-26)
Brugada syndrome (BrS) is a sudden death-predisposing genetic condition characterized electrocardiographically by ST segment elevation in the leads V(1)-V(3). Given the prominent role of the transient outward current (I(to)) in BrS pathogenesis, we hypothesized that rare gain-of-function mutations in KCND3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service