Skip to Content
Merck
All Photos(3)

Key Documents

AV31622

Sigma-Aldrich

Anti-SP7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Sp7 transcription factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SP7(121340)

General description

SP7 is a zinc-finger containing transcription factor that regulates BMP6-induced osteoblast differentiation.
Rabbit Anti-SP7 antibody recognizes bovine, human, mouse, and rat SP7.

Immunogen

Synthetic peptide directed towards the C terminal region of human SP7

Application

Rabbit Anti-SP7 antibody is suitable for use in western blot (0.125μg/ml) and immunohistochemical (4-8μg/ml) assays.

Biochem/physiol Actions

SP7 is a C2H2-type zinc finger transcription factor of the SP gene family and a putative master regulator of bone cell differentiation.

Sequence

Synthetic peptide located within the following region: RTHGEPGPGPPPSGPKELGEGRSTGEEEASQTPRPSASPATPEKAPGGSP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengchang Zhu et al.
Journal of cellular physiology, 227(6), 2677-2685 (2011-09-08)
The transcription factor osterix (Sp7) is essential for osteoblastogenesis and bone formation in mice. Genome wide association studies have demonstrated that osterix is associated with bone mineral density in humans; however, the molecular significance of osterix in human osteoblast differentiation
Xiao-Tao He et al.
Journal of cellular and molecular medicine, 22(2), 1302-1315 (2017-11-07)
Macrophages (Mφs) are involved in a variety of physiological and pathological events including wound healing and tissue regeneration, in which they play both positive and negative roles depending on their polarization state. In this study, we investigated the cellular behaviours
Ting Wang et al.
Phytotherapy research : PTR, 33(4), 1074-1083 (2019-02-16)
Osteoporosis is characterized by low bone mineral density and microarchitectural deterioration of bone tissue. N-(3-methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide (MBOC) is one of the macamides isolated from Maca (Lepidium meyenii Walp.), a cruciferous plant from the Andes of Peru. In this study, C3H/10T1/2 mesenchymal
Ting Wang et al.
Phytotherapy research : PTR, 33(4), 1074-1083 (2019-02-16)
Osteoporosis is characterized by low bone mineral density and microarchitectural deterioration of bone tissue. N-(3-methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide (MBOC) is one of the macamides isolated from Maca (Lepidium meyenii Walp.), a cruciferous plant from the Andes of Peru. In this study, C3H/10T1/2 mesenchymal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service