Skip to Content
Merck
All Photos(1)

Documents

94335

Sigma-Aldrich

Uridine 5′-diphosphoglucose disodium salt

≥98.0% (HPLC)

Synonym(s):

UDP-D-Glucose disodium salt, UDPG, UDP-glc, Uridine-diphosphate-glucose disodium salt, Uridine[5’]diphospho[1]-α-D-glucopyranose disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2Na2O17P2
CAS Number:
Molecular Weight:
610.27
Beilstein:
4290380
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

Saccharomyces cerevisiae

Assay

≥98.0% (HPLC)

form

powder

impurities

≤5% solvent
≤8.5% water

solubility

H2O: 50 mg/mL, clear

storage temp.

−20°C

SMILES string

[Na+].[Na+].OC[C@H]1O[C@H](OP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C15H24N2O17P2.2Na/c18-3-5-8(20)10(22)12(24)14(32-5)33-36(28,29)34-35(26,27)30-4-6-9(21)11(23)13(31-6)17-2-1-7(19)16-15(17)25;;/h1-2,5-6,8-14,18,20-24H,3-4H2,(H,26,27)(H,28,29)(H,16,19,25);;/q;2*+1/p-2/t5-,6-,8-,9-,10+,11-,12-,13-,14-;;/m1../s1

InChI key

PKJQEQVCYGYYMM-QBNUFUENSA-L

General description

Uridine5′-diphosphoglucose disodium salt is a nucleotide sugar consisting of a uracil base, ribose sugar, and two phosphate groups.

Application

UDP-glucose has been used:
  • as a substrate in the enzymatic production of glycosides and their detection by liquid chromatography-mass spectrometry.
  • as a substrate in the substrate screening and binding affinity measurements of the human CMP-Sia transporter.

Biochem/physiol Actions

UDP-glucose, acts as a precursor of UDP-galactose and UDP-glucuronate. In addition, it also aids in the biosynthesis of glucose-containing molecules such as oligosaccharides, polysaccharides, glycoproteins, and glycolipids in animals and some microorganisms . Several studies suggest that UDP-Glucose has the potential to act as a signaling molecule in plants beyond its classical metabolic functions . Uridine-5′-diphosphoglucose acts as an agonist to the purinergic receptor P2Y14 G protein-coupled receptor (GPCR) receptor, involved in the activation of dendritic cells and glial cells. It can also activate G protein-coupled receptor 17 (GPR17) thereby inducing oligodendrocyte differentiation.

Other Notes

Stimulates the reduction of methylcoenzyme M in cell-free extracts of Methanobacterium thermoautotrophicum

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Li et al.
Carbohydrate research, 446-447, 61-67 (2017-05-22)
Corylifol A, a member of the isoflavone subclass of isoflavonoids, has long been considered to have various biological activities. Here, we sought to synthesize corylifol A glucosides by the in vitro glucosylation reaction using the UDP-glycosyltransferase YjiC from Bacillus licheniformis DSM
Alexander Gutmann et al.
Organic & biomolecular chemistry, 15(37), 7917-7924 (2017-09-14)
Various bioactive natural products, like the aminocoumarin antibiotics novobiocin and coumermycin, exhibit an aromatic C-methyl group adjacent to a glycosylated phenolic hydroxyl group. Therefore, tailoring of basic phenolic scaffolds to contain the intricate C-methyl/O-glycosyl motif is of high interest for
Jo M Vanoevelen et al.
Journal of inherited metabolic disease, 41(1), 117-127 (2017-09-16)
Classic galactosemia is a genetic disorder of galactose metabolism, caused by severe deficiency of galactose-1-phosphate uridylyltransferase (GALT) enzyme activity due to mutations of the GALT gene. Its pathogenesis is still not fully elucidated, and a therapy that prevents chronic impairments
Ayako Ikegami et al.
Planta, 230(4), 841-855 (2009-07-31)
Fruits of persimmon (Diospyros kaki Thunb.) accumulate large amounts of proanthocyanidins (PAs) in the early stages of development. Astringent (A)-type fruits remain rich in soluble PAs even after they reach full-mature stage, whereas non-astringent (NA)-type fruits lose these compounds before
Sen Yin et al.
Plant physiology and biochemistry : PPB, 109, 536-548 (2016-11-12)
UDP-L-rhamnose (UDP-Rha) is an important sugar donor for the synthesis of rhamnose-containing compounds in plants. However, only a few enzymes and their encoding genes involved in UDP-Rha biosynthesis are available in plants. Here, two genes encoding rhamnose synthase (RhS) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service