Skip to Content
Merck
All Photos(1)

Documents

18854

Sigma-Aldrich

Kaempferol 7-O-β -D-glucopyranoside

≥90.0% (HPLC)

Synonym(s):

7-(β-D-Glucopyranosyloxy)-3,5-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Kaempferol 7-O-glucoside, Populnin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
Beilstein:
68205
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥90.0% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C(O)=C(Oc3c2)c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O11/c22-7-13-15(25)17(27)19(29)21(32-13)30-10-5-11(24)14-12(6-10)31-20(18(28)16(14)26)8-1-3-9(23)4-2-8/h1-6,13,15,17,19,21-25,27-29H,7H2/t13-,15-,17+,19-,21-/m1/s1

InChI key

YPWHZCPMOQGCDQ-HMGRVEAOSA-N

Biochem/physiol Actions

A flavonoid glycoside, found in plants such as aspen bark and leaves and poplar buds and bark, contains antimicrobial activities, and investigated for anticancer activities.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Isolation and quantification of kaempferol-7-O-glucoside and their antimicrobial screening of Cassia nodosa bunch.
Singh, D., et al.
Asian Journal of Biochemical and Pharmaceutical Research, 1, 6-16 (2011)
Yuan-Li Li et al.
Journal of ethnopharmacology, 113(1), 115-124 (2007-07-04)
The anticancer activity of eight crude extracts of Smilax china L. rhizome (SCR) against HeLa cells was assessed by MTT assay and clonogenic assay, the fraction rich in flavonoids had show good activity against HeLa cells. A bioassay-guided separation on
Angela Rubio Moraga et al.
BMC plant biology, 9, 109-109 (2009-08-22)
Flavonol glucosides constitute the second group of secondary metabolites that accumulate in Crocus sativus stigmas. To date there are no reports of functionally characterized flavonoid glucosyltransferases in C. sativus, despite the importance of these compounds as antioxidant agents. Moreover, their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service