Skip to Content
Merck
All Photos(2)

Key Documents

341428

Sigma-Aldrich

Ammonia solution

2.0 M in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH3
CAS Number:
Molecular Weight:
17.03
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

form

liquid

Quality Level

concentration

1.90-2.20 M (with NaOH, titration)
2.0 M in methanol

density

0.787 g/mL at 25 °C

SMILES string

N

InChI

1S/H3N/h1H3

InChI key

QGZKDVFQNNGYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Ammonia solution 25% can be used:
  • As a pH-adjusting agent to ensure beneficial alkaline conditions.
  • To prepare diammine silver(I) chloride complex by using silver chloride precipitate.
  • To prepare colloidal inks.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia A Wright et al.
Respiratory physiology & neurobiology, 184(3), 231-240 (2012-08-23)
In this review we pose the following seven questions related to ammonia and fish that represent gaps in our knowledge. 1. How is ammonia excretion linked to sodium uptake in freshwater fish? 2. How much does branchial ammonia excretion in
David A Stahl et al.
Annual review of microbiology, 66, 83-101 (2012-09-22)
The discovery of ammonia-oxidizing archaea (AOA), now generally recognized to exert primary control over ammonia oxidation in terrestrial, marine, and geothermal habitats, necessitates a reassessment of the nitrogen cycle. In particular, the unusually high affinity of marine and terrestrial AOA
Olivier Braissant et al.
Journal of inherited metabolic disease, 36(4), 595-612 (2012-10-31)
Hyperammonemia can be caused by various acquired or inherited disorders such as urea cycle defects. The brain is much more susceptible to the deleterious effects of ammonium in childhood than in adulthood. Hyperammonemia provokes irreversible damage to the developing central
Patrick M Erwin et al.
The ISME journal, 8(3), 575-588 (2013-10-25)
Ascidians are ecologically important components of marine ecosystems yet the ascidian microbiota remains largely unexplored beyond a few model species. We used 16S rRNA gene tag pyrosequencing to provide a comprehensive characterization of microbial symbionts in the tunic of 42
Hiroo Iwata et al.
Clinical pharmacokinetics, 43(4), 211-225 (2004-03-10)
We consider the pharmacokinetics of bioartificial livers (BALs) prepared using a hollow fibre module with respect to two key functions, detoxification and plasma protein supply, and present the results in a simple form. We then discuss the advantages and disadvantages

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service