Skip to Content
Merck
All Photos(1)

Documents

27221

Sigma-Aldrich

Acetic acid

glacial, puriss., 99-100%

Synonym(s):

Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.07 (vs air)

grade

puriss.

Assay

99-100%

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

technique(s)

mass spectrometry (MS): suitable

impurities

≤0.0005% heavy metals (as Pb)
≤0.005% non-volatile matter

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

Fe: ≤5 mg/kg

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetic acid is an organic carboxylic acid. It is a hygroscopic liquid having pungent odor. Its anhydrous form is commonly referred as glacial acetic acid. It forms dimers linked by hydrogen bonds in solution and gaseous phases. It is weakly acidic in nature.

Application

Acetic acid is widely employed as solvent for various oxidation reactions. It has been used in the analysis of synthetic peptides by electron transfer dissociation (ETD) tandem mass spectrometry (MS/MS). It may be used in the preparation of peroxyacetic acid (disinfectant), via reaction with perhydrol (30% hydrogen peroxide).

Other Notes

The article number 27221-6X1L will be discontinued. Please order the single bottle 27221-1L which is physically identical with the same exact specifications.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John E P Syka et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(26), 9528-9533 (2004-06-24)
Peptide sequence analysis using a combination of gas-phase ion/ion chemistry and tandem mass spectrometry (MS/MS) is demonstrated. Singly charged anthracene anions transfer an electron to multiply protonated peptides in a radio frequency quadrupole linear ion trap (QLT) and induce fragmentation
Eagleson M.
Concise Encyclopedia Chemistry, 5-5 (1994)
Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
Xianbao Deng et al.
The New phytologist, 201(4), 1469-1483 (2013-11-26)
• Chalcone synthase (CHS) is the key enzyme in the first committed step of the flavonoid biosynthetic pathway and catalyzes the stepwise condensation of 4-coumaroyl-CoA and malonyl-CoA to naringenin chalcone. In plants, CHS is often encoded by a small family

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service