Skip to Content
Merck
All Photos(3)

Key Documents

207810

Sigma-Aldrich

Potassium cyanide

ACS reagent, ≥96.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCN
CAS Number:
Molecular Weight:
65.12
Beilstein:
3593645
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥96.0%
grade:
ACS reagent
form:
granular
solubility:
water: soluble(lit.)

grade

ACS reagent

Quality Level

Assay

≥96.0%

form

granular

pH

11.5 (20 °C, 20 g/L)

mp

634 °C (lit.)

solubility

water: soluble(lit.)

anion traces

S2-: ≤0.003%
SCN-: passes test
chloride (Cl-): ≤0.5%
phosphate (PO43-): ≤0.005%
sulfate (SO42-): ≤0.04%

cation traces

Fe: ≤0.03%
Na: ≤0.5%
Pb: ≤2 ppm

SMILES string

[K]C#N

InChI

1S/CN.K/c1-2;

InChI key

YUZRZFQHUCKACF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium cyanide is a colorless hygroscopic compound. Its crystals exhibit cubic crystal system. It can be prepared by reacting hydrogen cyanide with potassium hydroxide. It participates in benzoin condensation reaction.

Application

Potassium cyanide may be employed in the following studies:
  • As a reducing agent in the colorimetric determination of amino acids.
  • Preparation of ferrocylacetonitrile, via reaction with methiodide of N,N-dimethylaminomethylferrocene.
  • Synthesis of O-acetylcyanohydrins.
  • Cyanation of aldimines via asymmetric Strecker-type reactions in the presence of chiral phase-transfer catalysts to form α-amino nitriles.
  • Metal-catalyzed addition of cyanide to 1,2-epoxides to form β-hydroxy nitriles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Strecker reaction of aldimines using aqueous potassium cyanide by phase-transfer catalysis of chiral quaternary ammonium salts with a tetranaphthyl backbone.
Ooi T, et al.
Journal of the American Chemical Society, 128(8), 2548-2549 (2006)
Easy direct stereo-and regioselective formation of ?-hydroxy nitriles by reaction of 1, 2-epoxides with potassium cyanide in the presence of metal salts.
Chini M, et al.
Tetrahedron Letters, 32(36), 4775-4778 (1991)
Eagleson M.
Concise Encyclopedia Chemistry, 18-18 (1994)
Rui Liu et al.
International journal of molecular sciences, 21(11) (2020-06-11)
Pentatricopeptide repeat (PPR) protein comprises a large family, participating in various aspects of organellar RNA metabolism in land plants. There are approximately 600 PPR proteins in maize, but the functions of many PPR proteins remain unknown. In this study, we
Reaction of the Methiodide of N, N-Dimethylaminomethylferrocene with Potassium Cyanide to Form Ferrocylacetonitrile1.
Lednicer D, et al.
The Journal of Organic Chemistry, 23(5), 653-655 (1958)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service