Skip to Content
Merck
All Photos(1)

Documents

Y0000017

Propofol for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,6-Diisopropylphenol, 2,6-Bis(1-methylethyl)phenol, 2,6-Bis(isopropyl)phenol, Propofol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2C6H3OH
CAS Number:
Molecular Weight:
178.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

5.6 mmHg ( 100 °C)

API family

propofol

manufacturer/tradename

EDQM

refractive index

n20/D 1.514 (lit.)

bp

256 °C/764 mmHg (lit.)

mp

18 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)c1cccc(C(C)C)c1O

InChI

1S/C12H18O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9,13H,1-4H3

InChI key

OLBCVFGFOZPWHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Propofol for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Suleman Khan et al.
Basic & clinical pharmacology & toxicology, 115(6), 565-570 (2014-06-04)
A variety of techniques have been developed to monitor the depth of anaesthesia. Propofol's pharmacokinetics and response vary greatly, which might be explained by genetic polymorphisms. We investigated the impact of genetic variations on dosage, anaesthetic depth and recovery after
Cynthia Ménard et al.
Radiology, 274(1), 181-191 (2014-09-10)
To determine if the integration of diagnostic magnetic resonance (MR) imaging and MR-guided biopsy would improve target delineation for focal salvage therapy in men with prostate cancer. Between September 2008 and March 2011, 30 men with biochemical failure after radiation
Jaime Acquaviva et al.
Molecular cancer research : MCR, 12(7), 1042-1054 (2014-05-03)
Activating mutations and/or overexpression of FGFR3 are common in bladder cancer, making FGFR3 an attractive therapeutic target in this disease. In addition, FGFR3 gene rearrangements have recently been described that define a unique subset of bladder tumors. Here, a selective
Philippe E Dubois et al.
European journal of anaesthesiology, 31(8), 404-410 (2013-07-26)
Disturbances in the thumb's movement interfere with the functioning of acceleromyography in many clinical settings. The short and light (SL) train-of-four (TOF)-Tube is a new version of a rigid tubular device that was designed to protect the thumb from external
Eran A Mukamel et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(3), 839-845 (2014-01-17)
Rhythmic oscillations shape cortical dynamics during active behavior, sleep, and general anesthesia. Cross-frequency phase-amplitude coupling is a prominent feature of cortical oscillations, but its role in organizing conscious and unconscious brain states is poorly understood. Using high-density EEG and intracranial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service