Skip to Content
Merck
All Photos(1)

Key Documents

T2900000

Tyrosine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Tyrosine, (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
Beilstein:
392441
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tyrosine

manufacturer/tradename

EDQM

mp

>300 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1

InChI key

OUYCCCASQSFEME-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tyrosine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Amino acid precursor of dopamine and other catecholamines
Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philippe Menasché et al.
European heart journal, 36(12), 743-750 (2014-05-20)
There is now compelling evidence that cells committed to a cardiac lineage are most effective for improving the function of infarcted hearts. This has been confirmed by our pre-clinical studies entailing transplantation of human embryonic stem cell (hESC)-derived cardiac progenitors
Dana Shani Galili et al.
Current biology : CB, 24(15), 1712-1722 (2014-07-22)
Drosophila learn to avoid odors that are paired with aversive stimuli. Electric shock is a potent aversive stimulus that acts via dopamine neurons to elicit avoidance of the associated odor. While dopamine signaling has been demonstrated to mediate olfactory electric
Martin Schlumberger et al.
The New England journal of medicine, 372(7), 621-630 (2015-02-12)
Lenvatinib, an oral inhibitor of vascular endothelial growth factor receptors 1, 2, and 3, fibroblast growth factor receptors 1 through 4, platelet-derived growth factor receptor α, RET, and KIT, showed clinical activity in a phase 2 study involving patients with
Z Xiong et al.
Stem cell research, 12(2), 387-399 (2014-01-01)
An essential step for therapeutic and research applications of stem cells is their ability to differentiate into specific cell types. Neuronal cells are of great interest for medical treatment of neurodegenerative diseases and traumatic injuries of central nervous system (CNS)
Elpida Tsika et al.
Human molecular genetics, 23(17), 4621-4638 (2014-04-18)
Mutations in the vacuolar protein sorting 35 homolog (VPS35) gene at the PARK17 locus, encoding a key component of the retromer complex, were recently identified as a new cause of late-onset, autosomal dominant Parkinson's disease (PD). Here we explore the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service