Skip to Content
Merck
All Photos(1)

Documents

QC1380

Supelco

Formaldehyde in Water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 20 mL

manufacturer/tradename

RTC QCO-417

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
food and beverages

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa M Lamb et al.
PloS one, 6(11), e27149-e27149 (2011-11-17)
MAPK signal transduction pathways are important regulators of stress responses, cellular growth, and differentiation. In Neurospora, the circadian clock controls rhythms in phosphorylation of the p38-like MAPK (OS-2); however, the mechanism for this regulation is not known. We show that
Shiuh-Dih Chou et al.
Molecular immunology, 45(4), 971-980 (2007-09-14)
We have previously reported that Major Histocompatibility Complex (MHC) class II can be induced by histone deacetylase inhibitors (HDACi) in the absence of class II transactivator (CIITA). Here we characterized the histone modifications associated with the CIITA-dependent (IFN-gamma induced) and
Katy R Newton et al.
Immunology, 125(4), 469-479 (2008-05-31)
Dendritic cells (DCs) play a key role in the type and course of an immune response. The manipulation of human DCs to produce therapeutic agents by transduction with viral vectors is a growing area of research. We present an investigation
Hongpeng He et al.
PloS one, 6(7), e22088-e22088 (2011-07-27)
Histones constitute a type of essential nuclear proteins important for chromatin structure and functions. The expression of major histones is strictly confined to the S phase of a cell cycle and tightly coupled to DNA replication. With RT-qPCR and ChIP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service