Skip to Content
Merck
All Photos(1)

Key Documents

PHR1079

Supelco

Tobramycin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Nebramycin Factor 6, O-[3-Amino-3-deoxy-α-D-glucopyranosyl-(1→6)]-O-[2,6-diamino-2,3,6-trideoxy-α-D-ribohexopyranosyl-(1→4)]-2-deoxy-D-streptamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H37N5O9
CAS Number:
Molecular Weight:
467.51
Beilstein:
1357507
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 333
traceable to Ph. Eur. T1500000
traceable to USP 1667508

API family

tobramycin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@@H](N)[C@H]3O)[C@H]2O)[C@H](N)C[C@@H]1O

InChI

1S/C18H37N5O9/c19-3-9-8(25)2-7(22)17(29-9)31-15-5(20)1-6(21)16(14(15)28)32-18-13(27)11(23)12(26)10(4-24)30-18/h5-18,24-28H,1-4,19-23H2/t5-,6+,7+,8-,9+,10+,11+,12+,13+,14-,15+,16-,17+,18+/m0/s1

InChI key

NLVFBUXFDBBNBW-SNGYORCQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards. Tobramycin is an aminoglycoside antibiotic effective in treating infections caused by Gram negative bacteria.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Tobramycin may be used as a pharmaceutical reference standard for the determination of the analyte in drug dosage forms by high performance liquid chromatography.

Biochem/physiol Actions

Tobramycin is an aminoglycoside.
Mode of Action: Binds to 70S ribosomal subunit; inhibits translocation; elicits miscoding.
Spectrum of Activity: Gram negative bacteria. Not effective against Enterococci.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA5713 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple and rapid high-performance liquid chromatographic method for determining tobramycin in pharmaceutical formulations by direct UV detection
Ruckmani K, et al.
Pharmaceutical Methods, 2(2), 117-123 (2011)
Determination of aminoglycosides in pharmaceutical formulations-II. High-performance liquid chromatography
Fabre H, et al.
Journal of Pharmaceutical and Biomedical Analysis, 7(12), 1711-1718 (1989)
A liquid-chromatographic method for the determination of tobramycin
Dash AK and Suryanarayanan R
Journal of Pharmaceutical and Biomedical Analysis, 9(3), 237-245 (1991)
Development of molecular imprinted nanosensor for determination of tobramycin in pharmaceuticals and foods
Yola M, et al.
Talanta, 120(2), 318-324 (2014)
Stefanie Hennig et al.
Clinical pharmacokinetics, 52(4), 289-301 (2013-02-20)
While several studies have examined the pharmacokinetics of tobramycin in patients with cystic fibrosis (CF), there is no consensus on whether they differ in patients with and without CF. The objectives of this study were to identify covariates which explain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service