Skip to Content
Merck
All Photos(1)

Key Documents

I0361000

Ipratropium bromide impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(1R,3r,5S,8s)-3-[[(2RS)-3-Hydroxy-2-phenylpropanoyl]oxy]-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30BrNO3
CAS Number:
Molecular Weight:
412.36
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ipratropium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ipratropium bromide impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary T Ferguson et al.
International journal of chronic obstructive pulmonary disease, 8, 139-150 (2013-05-10)
Ipratropium bromide/albuterol Respimat inhaler (CVT-R) was developed as an environmentally friendly alternative to ipratropium bromide/albuterol metered-dose inhaler (CVT-MDI), which uses a chlorofluorocarbon propellant. The objective of this study was to evaluate patient satisfaction, device usage, and long-term safety of CVT-R
Yuko Kawasumi et al.
Clinical therapeutics, 35(4), 523-531 (2013-03-16)
In the treatment of chronic obstructive pulmonary disease (COPD), tiotropium bromide has a longer duration of action than ipratropium bromide; however, tiotropium bromide is a more expensive alternative treatment. At issue is whether tiotropium reduces the risk for hospital readmissions
Sanja Juric et al.
Xenobiotica; the fate of foreign compounds in biological systems, 43(9), 785-791 (2013-04-11)
Human hepatocytes that had been cold-preserved in SureTran(TM) matrix (Abcellute Ltd, Cardiff, UK) were used for studies on cell viability, cytochrome P450 (CYP) 3A4, 2B6 and 1A2 induction and hepatic drug transporters. It has recently been shown that basal CYP
Predictors of adherence to inhaled medications.
Ami Schattner
Journal of general internal medicine, 28(6), 758-758 (2013-03-05)
Predictors of adherence to inhaled medications.
John C Huetsch et al.
Journal of general internal medicine, 28(6), 759-759 (2013-03-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service