Skip to Content
Merck
All Photos(1)

Documents

E1700000

Estrone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,3,5(10)-Estratrien-3-ol-17-one, 3-Hydroxy-1,3,5(10)-estratrien-17-one, Folliculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
Beilstein:
1915077
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

estrone

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

258-260 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CCC2=O

InChI

1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1

InChI key

DNXHEGUUPJUMQT-CBZIJGRNSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Estrone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Emma M Ferguson et al.
Water research, 47(4), 1604-1615 (2013-01-12)
Estuaries are often the final repositories for aquatic pollutants but how estuarine hydrology influences the availability of marine- and freshwater-derived pollutants is not well understood, particularly for micro-pollutants such as endocrine disrupting chemicals. To address this knowledge gap, this study
Shanshan Zhao et al.
Breast cancer research : BCR, 16(2), R30-R30 (2014-03-29)
Paradoxically, a breast cancer risk reduction with conjugated equine estrogens (CEE) and a risk elevation with CEE plus medroxyprogesterone acetate (CEE + MPA) were observed in the Women's Health Initiative (WHI) randomized controlled trials. The effects of hormone therapy on serum sex
Yoko Urata et al.
The Journal of clinical endocrinology and metabolism, 98(4), 1583-1590 (2013-03-02)
Endometriosis is a chronic inflammatory disease in which immune response and production of estrogen in endometriotic tissues are involved in the development of the disease. Prostaglandin E2 (PGE2) stimulates aromatase (P450arom) expression in endometrioma stromal cells (ESCs) and increases the
B Zumoff et al.
Arteriosclerosis (Dallas, Tex.), 2(1), 58-67 (1982-01-01)
Plasma concentrations and urinary excretions of various hormones and hormone metabolites were measured in four groups. Group 1 was composed of 13 men with prior myocardial infarction; Group 2 contained 35 clinically normal men; Group 3 consisted of 44 men
Juilee Rege et al.
The Journal of clinical endocrinology and metabolism, 98(3), 1182-1188 (2013-02-07)
A broad analysis of adrenal gland-derived 19-carbon (C19) steroids has not been reported. This is the first study that uses liquid chromatography-tandem mass spectrometry to quantify 9 C19 steroids (androgens and their precursors), estrone, and estradiol in the adrenal vein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service