Skip to Content
Merck
All Photos(3)

Key Documents

A9085

Sigma-Aldrich

Bovine Serum Albumin

lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

bovine

Quality Level

Assay

≥96% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

poly bottle of

origin

USA origin

concentration

<25 ng/mg IgG

technique(s)

flow cytometry: suitable
immunofluorescence: suitable

impurities

IgG free
≤25 ng/mg protein IgG

pH

7

solubility

water: soluble (40 mg/ml)

UniProt accession no.

foreign activity

Blue tongue virus, none detected
Vesicular Statitis virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine serum albumin (BSA) is a 66 kDa protein consisting of three domains with two subdomains under each. It is a α-helical, globular and non-glycosylated protein. BSA belongs to the serum albumins family. It has 17-disulfide bonds.

Application

Bovine Serum Albumin has been used in:
  • the washing of the rabbit tissue sections in immunofluorescence microscopy
  • incubation of red blood cells (RBC) for flow cytometry analysis
  • as a component of Hanks′ balanced salt solution with Ca2+ and Mg2+ (HBSS++) for eosin-5-maleimide staining and RBC washing in magnetic circular dichroism measurements

Biochem/physiol Actions

Bovine Serum Albumin (BSA) is a transporter for drugs, hormones and fatty acids. It is used as a blocking agent in enzyme linked immunosorbent assay (ELISA) for preventing non-specific binding of antigens and antibodies to the microtiter plates. It is a crucial component of the cell culture media and favors embryonic stem cells (hESC) differentiation. BSA shares structural features with human serum albumin. Its high solubility, low cost and purity and interaction with surfactant is exploited in cosmetic and pharmaceutical industry.
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

  • Essentially IgG free (immunoglobulin-free)
  • Heat shock fractionated

Preparation Note

Prepared using heat shock fractionation.
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prenatal Exposure to Vinclozolin Disrupts Selective Aspects of the Gonadotrophin-Releasing Hormone Neuronal System of the Rabbit
Wadas BC, et al.
Journal of Neuroendocrinology, 22(6), 518-526 (2010)
C4d Deposits on the Surface of Red Blood Cells in Trauma Patients and Interferes with their Function
Muroya T, et al.
Critical Care Medicine, 42(5), e364-e364 (2014)
Javier Encinar Del Dedo et al.
Developmental cell, 43(5), 588-602 (2017-11-28)
Oxysterol binding protein-related proteins (ORPs) are conserved lipid binding polypeptides, enriched at ER contacts sites. ORPs promote non-vesicular lipid transport and work as lipid sensors in the context of many cellular tasks, but the determinants of their distinct localization and
Binding of fatty acid amide amphiphiles to bovine serum albumin: role of amide hydrogen bonding
Ghosh S and Dey J
The Journal of Physical Chemistry B, 119(25), 7804-7815 (2015)
Albumin and mammalian cell culture: implications for biotechnology applications
Francis GL
Cytotechnology, 62(1), 1-16 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service