Skip to Content
Merck
All Photos(1)

Key Documents

82281

Sigma-Aldrich

1,2-Propanediol

tested according to Ph. Eur.

Synonym(s):

1,2-PDO, Propylenglycolum, Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

form

viscous liquid

autoignition temp.

779 °F

expl. lim.

12.5 %

refractive index

n20/D 1.432 (lit.)

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gai Ling Li et al.
Journal of controlled release : official journal of the Controlled Release Society, 84(1-2), 49-57 (2002-10-26)
To achieve a therapeutical effect of the anti-Parkinson's drug R-apomorphine via iontophoresis delivery, enhancement strategies in vitro were explored using three structurally related enhancers, lauric acid (LA), dodecyltrimethylammonium bromide (DTAB) and Laureth-3 oxyethylene ether (C(12)EO(3)). Human stratum corneum and shed
R S Rajasenan et al.
Toxicology and applied pharmacology, 135(1), 89-99 (1995-11-01)
Engrafting components of human immune systems in severe combined immunodeficient (SCID) mice has been utilized to investigate the pathogenesis of several human autoimmune diseases and may provide a model for studying idiosyncratic drug toxicity. The purpose of this investigation was
G A Reichard et al.
Diabetes, 35(6), 668-674 (1986-06-01)
Plasma acetone turnover rates were measured with the primed continuous infusion of 2-[14C]acetone in patients with moderate to severe diabetic ketoacidosis. Plasma acetone turnover rates ranged from 1.52 to 15.9 mumol X kg-1 X min-1 (108-1038 mumol X 1.73 m-2
Rong-Yao Wang et al.
Physical chemistry chemical physics : PCCP, 15(9), 3313-3319 (2013-01-31)
Understanding the role of kinetics in fiber network microstructure formation is of considerable importance in engineering gel materials to achieve their optimized performances/functionalities. In this work, we present a new approach for kinetic-structure analysis for fibrous gel materials. In this
Maria Pina Rosato et al.
Theriogenology, 79(3), 508-516 (2012-12-12)
This study was designed to improve current freezing protocols for rabbit sperm by examining: (1) the toxicity of different permeable cryoprotectants (CPAs) used for standard vapor freezing (conventional freezing); (2) the feasibility of ultrarapid nonequilibrium freezing (vitrification) of sperm in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service