Skip to Content
Merck
All Photos(1)

Documents

76631

Supelco

Methyl salicylate

analytical standard

Synonym(s):

2-Hydroxybenzoic acid methyl ester, Methyl 2-hydroxybenzoate, Oil of wintergreen, Wintergreen oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
971516
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.26 (vs air)

vapor pressure

1 mmHg ( 54 °C)

Assay

≥99.0% (GC)

autoignition temp.

847 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.536 (lit.)
n20/D 1.536-1.539

bp

222 °C (lit.)

mp

−8-−7 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COC(=O)c1ccccc1O

InChI

1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

OSWPMRLSEDHDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl Salicylate is a methyl ester of salicylic acid, widely used as phytohormones and defensive signaling compounds. It is most commonly used in topical products as a rubefacients.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Methyl Salicylate may be used as an analytical reference standard for the quantification of the analyte in Ulmus pumila leaves using gas chromatography coupled to mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Simultaneous determination of salicylic acid, jasmonic acid, methyl salicylate, and methyl jasmonate from Ulmus pumila leaves by GC-MS
Huang H-Z, et al.
International Journal of Analytical Chemistry (2015)
The Essence of Analgesia and Analgesics (2010)
Agata Bartyzel et al.
Analytica chimica acta, 707(1-2), 204-209 (2011-10-27)
N,N'-bis-(α-methylsalicylidene)-2,2-dimethyl-1,3-propanediimine (SBTD) modified silica gel was prepared and used as sorbent for solid phase extraction of Cr(III) ions from aqueous solution. This sorbent showed a high sorption affinity for Cr(III) while recovery of Cr(VI) was very low. The analyte ion
Michiel van Wijk et al.
PloS one, 6(7), e21742-e21742 (2011-07-19)
Insect herbivory induces plant odors that attract herbivores' natural enemies. Assuming this attraction emerges from individual compounds, genetic control over odor emission of crops may provide a rationale for manipulating the distribution of predators used for pest control. However, studies
Nadir Erbilgin et al.
Tree physiology, 32(8), 946-957 (2012-06-05)
Coniferous trees have both constitutive and inducible defences that deter or kill herbivores and pathogens. We investigated constitutive and induced monoterpene responses of jack pine (Pinus banksiana Lamb.) to a number of damage types: a fungal associate of the mountain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service