Skip to Content
Merck
All Photos(1)

Documents

68878

Supelco

(+)-Borneol

analytical standard

Synonym(s):

endo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2038056
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)

optical activity

[α]20/D 37.0±2.0°, c = 5 in ethanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

206-209 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@@](C)([C@@H](O)C1)C2(C)C

InChI

1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8+,10+/m1/s1

InChI key

DTGKSKDOIYIVQL-WEDXCCLWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Borneol, a bicyclic monoterpenoid alcohol which is a major constituent of medicinal plants is grouped under the volatile organic compounds (VOCs) family of chemicals. It is employed as analgesic and anesthetic in traditional Chinese and Japanese medicines.

Application

(+)-Borneol has been reported exert highly efficient positive modulating gamma-aminobutyric action (GABA) action at human recombinant α1β2γ2L GABAA receptors.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: angelica cinnamomum curcuma echinacea salvia thymus valeriana zingiber

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The enhancing effect of synthetical borneol on the absorption of tetramethylpyrazine phosphate in mouse.
Yan-Yu X, et al.
International Journal of Pharmaceutics, 337(1-2), 74-79 (2007)
Supercritical carbon dioxide extraction of volatiles from spices: comparison with simultaneous distillation-extraction.
Diaz-Maroto MC, et al.
Journal of Chromatography A, 947(1), 23-29 (2002)
(+)-And (-)-borneol: efficacious positive modulators of GABA action at human recombinant a 1 ? 2 ? 2L GABA A receptors.
Granger RE, et al.
Biochemical Pharmacology, 69(7), 1101-1111 (2005)
Jianyu Su et al.
Journal of food science, 77(6), C658-C664 (2012-05-16)
The aims of this study were to optimize the preparation conditions of natural borneol/β-cyclodextrin (NB/β-CD) inclusion complex by ultrasound method, and to investigate its improvement of stability and solubility. The complex was characterized by different various spectroscopic techniques including Fourier
Xiao-Hong Wei et al.
Microcirculation (New York, N.Y. : 1994), 20(1), 17-29 (2012-08-24)
The present study was designed to evaluate whether CP was beneficial in alleviating myocardial fibrosis following I/R injury. Sprague-Dawley rats were subjected to 30 minutes occlusion of the LADCA, followed by reperfusion. CP (0.4 or 0.8 g/kg) was daily administered starting from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service