Skip to Content
Merck
All Photos(1)

Documents

45393

Supelco

Chlorpropham

PESTANAL®, analytical standard

Synonym(s):

Chloro-ICP, Chloropropham, Isopropyl N-(3-chlorophenyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H12NO2Cl
CAS Number:
Molecular Weight:
213.66
Beilstein:
2211397
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)OC(=O)Nc1cccc(Cl)c1

InChI

1S/C10H12ClNO2/c1-7(2)14-10(13)12-9-5-3-4-8(11)6-9/h3-7H,1-2H3,(H,12,13)

InChI key

CWJSHJJYOPWUGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ona Sakaliene et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 44(1), 1-6 (2008-12-18)
Chlorpropham (isopropyl 3-chlorocarbanilate) is a pesticide used to control sprouting of potatoes during long-term storage. The objective of the present study was to establish the total chlorpropham residue balance (residues in unwashed and washed whole tubers, peeled tubers, peels, boiled
Y Nakagawa et al.
Xenobiotica; the fate of foreign compounds in biological systems, 34(3), 257-272 (2004-06-19)
1: The metabolism and action of chlorpropham (isopropyl N-(3-chlorophenyl)carbamate; CIPC, a post-harvest agent) were studied in freshly isolated rat hepatocytes, and the oestrogen-like activity of CIPC and its metabolites was assessed by in vitro assays. The exposure of hepatocyte suspensions
Junichi Eto et al.
FEBS letters, 550(1-3), 179-184 (2003-08-26)
An anti-chlorpropham single-chain variable-fragment (scFv) gene was introduced into Arabidopsis in a manner to express the antibody fragment in each of four different subcellular compartments. The accumulation of scFv in transgenic plants was detected by targeting the fragment in the
Margaret J Smith et al.
Journal of hazardous materials, 246-247, 154-162 (2013-01-09)
DFT calculations were performed on the thermal reactions of chlorpropham 1, a carbamate pesticide and plant growth regulator frequently used in the storage of potatoes. At the conditions normally used in applying 1 (injection of a methanolic solution of 1
Helen Ferrier et al.
Food additives and contaminants, 23(6), 601-615 (2006-06-13)
The assessment of consumer exposure to pesticides is an important part of pesticide regulation. Probabilistic modelling allows analysis of uncertainty and variability in risk assessments. The output of any assessment will be influenced by the characteristics and uncertainty of the

Protocols

-methylcarbamate 10 μg/mL; Diuron; Propham; Siduron; Methiocarb, analytical standard; Linuron 10 μg/mL; Swep 10 μg/mL; Chlorpropham 10 μg/mL; Barban; Neburon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service