Skip to Content
Merck
All Photos(1)

Documents

39496

Supelco

L-Methionine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(S)-2-Amino-4-(methylmercapto)butyric acid, L-2-Amino-4-(methylthio)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
149.21
Beilstein:
1722294
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

284 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

20-25°C

SMILES string

CSCC[C@H](N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1

InChI key

FFEARJCKVFRZRR-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald O Ball et al.
The Journal of nutrition, 136(6 Suppl), 1682S-1693S (2006-05-17)
Sulfur amino acid metabolism has been receiving increased attention because of the link to chronic diseases such as cardiovascular disease, Alzheimer's disease, and diabetes. In addition, the role of cysteine and optimal intakes for physiological substrates such as glutathione are
Alex G Harrison
Journal of the American Society for Mass Spectrometry, 24(10), 1555-1564 (2013-08-15)
The fragmentation reactions of the MH(+) ions as well as the b7, a7, and a7* ions derived therefrom have been studied in detail for the octapeptides MAAAAAAA, AAMAAAAA, AAAAMAAA, and AAAAAAMA. Ionization was by electrospray using a QqToF mass spectrometer
Andor W J M Glaudemans et al.
European journal of nuclear medicine and molecular imaging, 40(4), 615-635 (2012-12-13)
(11)C-methionine (MET) is the most popular amino acid tracer used in PET imaging of brain tumours. Because of its characteristics, MET PET provides a high detection rate of brain tumours and good lesion delineation. This review focuses on the role
Camilla Schalin-Jäntti et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(5), 739-747 (2013-04-05)
All patients with primary hyperparathyroidism should undergo localization studies before reoperation, but it is not known which method is most accurate. The purpose of this prospective study was to compare the performance of planar scintigraphy with (123)I/(99m)Tc-sestamibi, (99m)Tc-sestamibi SPECT (SPECT/CT)
Olga Z Karicheva et al.
Nucleic acids research, 39(18), 8173-8186 (2011-07-05)
Mutations in human mitochondrial DNA are often associated with incurable human neuromuscular diseases. Among these mutations, an important number have been identified in tRNA genes, including 29 in the gene MT-TL1 coding for the tRNA(Leu(UUR)). The m.3243A>G mutation was described

Articles

Separation of L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material

Separation of L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material

Separation of L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material

Separation of L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service