Skip to Content
Merck
All Photos(1)

Documents

33703

Supelco

Ofloxacin

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20FN3O4
CAS Number:
Molecular Weight:
361.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CC1COc2c(N3CCN(C)CC3)c(F)cc4C(=O)C(=CN1c24)C(O)=O

InChI

1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)

InChI key

GSDSWSVVBLHKDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly M Khomtchouk et al.
Science advances, 6(33), eabc1828-eabc1828 (2020-08-28)
Chronic suppurative otitis media (CSOM) is a neglected pediatric disease affecting 330 million worldwide for which no new drugs have been introduced for over a decade. We developed a mouse model with utility in preclinical drug evaluation and antimicrobial discovery.
S Güniz Küçükgüzel et al.
European journal of medicinal chemistry, 42(7), 893-901 (2007-04-10)
Three novel series of 2',4'-difluoro-4-hydroxybiphenyl-3-carboxylic acid derivatives namely 4-substituted-1,2,4-triazoline-3-thiones (4a-g); 2-substituted-1,3,4-thiadiazoles (5a-g) and 2-substituted-1,3,4-oxadiazoles (6a-g) have been synthesized. Twenty-one of the newly synthesized compounds were tested against various bacteria, fungi, yeast species and virus. In addition, we have replaced the
Duy An Duong et al.
Antimicrobial agents and chemotherapy, 53(11), 4835-4839 (2009-09-02)
Consecutive fluoroquinolone (FQ)-resistant isolates (n = 109) identified at the Pham Ngoc Thach Hospital for Tuberculosis, Ho Chi Minh City, Vietnam, were sequenced in the quinolone resistance-determining regions of the gyrA and gyrB genes and typed by large sequence polymorphism
Tarja Pohjanvirta et al.
Pathogens (Basel, Switzerland), 9(10) (2020-10-04)
Mycoplasma bovis is an important bovine pathogen. Artificial insemination (AI) using contaminated semen can introduce the agent into a naïve herd. Antibiotics, most often gentamycin, tylosin, lincomycin, spectinomycin (GTLS) combination are added to semen extender to prevent transmission of pathogenic
Andrea Von Groll et al.
Antimicrobial agents and chemotherapy, 53(10), 4498-4500 (2009-08-19)
This study evaluated cross-resistance of Mycobacterium tuberculosis strains to ofloxacin, moxifloxacin, and gatifloxacin and investigated the presence of mutations in gyrA and gyrB. Fluoroquinolone susceptibilities were determined for 41 M. tuberculosis strains by the proportion method on 7H11, and MICs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service