Skip to Content
Merck
All Photos(1)

Documents

11669940910

Roche

DNA Molecular Weight Marker VII, DIG-labeled

greener alternative

Synonym(s):

DNA marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335

form

solution

Quality Level

packaging

pkg of 500 μL (10 μg/ml)

manufacturer/tradename

Roche

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

10 μg/mL

color

colorless

solubility

water: miscible

greener alternative category

storage temp.

−20°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical. The DIG System was established as a sensitive and cost-effective alternative to using radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.
DNA Molecular Weight Marker VII, DIG-labeled is a ready-to-use solution in 10mM Tris-HCl, 1mM EDTA, pH 8.0. The fragment mixture is prepared by cleavage of SPP1 DNA with EcoRI. the size ranges from 0.081 to 8.57kbp.

Application

DNA Molecular Weight Marker VII, DIG-labeled, has been used as a size standard in Southern blot analysis for nucleic acid labeling and detection.

Sequence

The mixture contains 17 fragments wit the following base pair lengths: 81, 359, 492, 710, 718, 992, 1164, 1482, 1515, 1882, 1953, 2799, 3639, 4899, 6106, 7427, and 8576 bp.

Note: Fragment lengths are derived from computer analysis of the DNA sequence. Depending on the size range of the marker, the smallest fragments will only be visible on overloaded gels.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fei Gao et al.
Molecular genetics & genomic medicine, 8(6), e1236-e1236 (2020-04-14)
Fragile X syndrome (FXS) is the most common inherited form of intellectual disability caused by a CGG repeat expansion in the 5' untranslated region of the FMR1 gene. When the number of repeats exceeds 200, the gene becomes hypermethylated and
Hui Xi et al.
Molecular genetics & genomic medicine, 9(7), e1711-e1711 (2021-06-01)
Fragile X syndrome (FXS) is the most common inherited form of intellectual disability. Prenatal screening of FXS allows for early identification and intervention. The present study explored the feasibility of FXS carrier screening during prenatal diagnosis for those who were
Wen Huang et al.
Molecular genetics & genomic medicine, 3(3), 172-181 (2015-06-02)
Fragile X syndrome is mainly caused by a CGG repeat expansion within the 5' UTR of the fragile X mental retardation 1 (FMR1) gene. Previous analyses of the FMR1 CGG repeat patterns and flanking haplotypes in Caucasians and African Americans
Kathryn M McCulloch et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(37), 11547-11552 (2015-08-05)
Orthosomycins are oligosaccharide antibiotics that include avilamycin, everninomicin, and hygromycin B and are hallmarked by a rigidifying interglycosidic spirocyclic ortho-δ-lactone (orthoester) linkage between at least one pair of carbohydrates. A subset of orthosomycins additionally contain a carbohydrate capped by a

Articles

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service