Skip to Content
Merck
All Photos(1)

Key Documents

11669915001

Roche

Cell Proliferation ELISA, BrdU (chemiluminescent)

Synonym(s):

BrdU, cell proliferation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158

usage

sufficient for ≤1,000 tests

Quality Level

manufacturer/tradename

Roche

technique(s)

ELISA: suitable

detection method

chemiluminescent

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

Chemiluminescent immunoassay for the quantification of cell proliferation, based on the measurement of BrdU incorporation during DNA synthesis:  a nonradioactive alternative to the [3H]-thymidine incorporation assay.

Specificity

The antibody conjugate reacts with the thymidine analogue 5-bromo-2′-deoxyuridine (BrdU) and with BrdU incorporated into DNA. For binding to BrdU incorporated into the DNA, the BrdU-labeled DNA has to be denatured. The antibody does not cross-react with any endogenous cellular components such as thymidine, uridine, or DNA.

Application

Cell Proliferation ELISA, BrdU (chemiluminescent) has been used:
  • for bromodeoxyuridine (BrdU) proliferation assay in ReN and primary astrocytes
  • to test the effect on imatinib on the proliferation of neuroblastoma cells using BrdU incorporation assay
  • to check the effect of palbociclib on cell cycle inhibition using BrdU incorporation in pancreatic patient-derived primary cell lines

The Cell Proliferation ELISA, BrdU (chemiluminescent) belongs to the second, improved generation of kits for measuring DNA synthesis. It is a precise, fast, and simple nonradioactive alternative to quantitate cell proliferation based on the measurement of BrdU incorporation during DNA synthesis in replicating (cycling) cells. Thus, the Cell Proliferation ELISA can be used in many different in vitro cell systems. For example:
  • Detection and quantification of cell proliferation induced by growth factors and cytokines
  • Determination of the inhibitory or stimulatory effects of various compounds on cell proliferation in environmental and biomedical research, and in the food, cosmetic, and pharmaceutical industries
  • Measurement of the immunoreactivity of lymphocytes, stimulated by mitogens or antigens
  • Analysis of the chemosensitivity of tumor cells to different cytostatic drugs in medical research

Packaging

1 kit containing 7 components.

Components

  1. BrdU Labeling Reagent, 1,000x conc
  2. FixDenat
  3. Anti-BrdU-POD 4. Antibody Dilution Solution
  4. Washing Buffer PBS, 10x conc.
  5. Substrate Component A
  6. Substrate Component B

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • BrdU Labeling Reagent

  • FixDenat ready-to-use

  • Anti-BrdU-peroxidase antibody

  • Antibody Dilution Solution ready-to-use

  • ready-to-use 10x concentrated

  • Substrate Component A

  • Substrate Component B

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Feng Wang et al.
Molecular medicine reports, 12(4), 5917-5923 (2015-08-25)
Epirubicin, an anthracycline derivative, is one of the main line treatments for brain tumors. The aim of the present study was to verify that epirubicin alters the growth and morphological characteristics of U‑87 glioma cells. In the present study, the
Valerio Ciccone et al.
Oncotarget, 9(17), 13353-13365 (2018-03-24)
Nitric oxide (NO) exerts conflicting effect on tumor growth and progression, depending on its concentration. We aimed to characterize the anti-cancer activity of a new NO donor, Ni(SalPipNONO) belonging to the class of metal-nonoates, in epithelial derived tumor cells, finally
Diego S D'Astolfo et al.
Cell, 161(3), 674-690 (2015-04-25)
Modulation of protein function is used to intervene in cellular processes but is often done indirectly by means of introducing DNA or mRNA encoding the effector protein. Thus far, direct intracellular delivery of proteins has remained challenging. We developed a
Juliann E Kosovec et al.
Oncotarget, 8(59), 100421-100432 (2017-12-17)
Esophageal adenocarcinoma (EAC) is a deadly disease with limited therapeutic options. In the present study, we determined the preclinical efficacy of CDK4/6 inhibitor abemaciclib for treatment of EAC. In vitro, apoptosis, proliferation, and pathway regulation were evaluated in OE19, OE33
Tino Schneider et al.
Glycobiology, 25(8), 812-824 (2015-04-17)
The present study demonstrates that fucose-containing sulfated polysaccharides (FCSP) from brown algae interfere with the CXCL12/CXCR4 axis in human Burkitt's lymphoma cells by binding CXCL12 and thereby blocking both CXCL12-induced CXCR4 receptor activation and downstream effects like migration and secretion

Articles

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service