Skip to Content
MilliporeSigma

Skip To

PP40

IgG, Goat

Goat IgG Polyclonal Antibody control validated for use in ELISA.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Check Cart for Availability
$392.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Technique(s):
ELISA: suitable
Species reactivity:
-
Application:
ELISA
Citations:
11

$392.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

antibody form

purified immunoglobulin

manufacturer/tradename

Chemicon®

concentration

4.59 mg/mL

technique(s)

ELISA: suitable

shipped in

dry ice

target post-translational modification

unmodified

General description

Source: Normal goat serum.
Purity: 95%, purified by salt fractionation, ion exchange chromatography. Single band exhibited when run against anti-species specific whole serum.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Physical form

Format: Purified
Purified immunoglobulin. Liquid filtered through 0.45 μm Swinex, no preservatives.

Preparation Note

Store -20°C for up to 12 months. Avoid repeated freeze thaw cycle.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I9140I5256A3688
biological source

goat

biological source

-

biological source

-

biological source

goat

antibody form

purified immunoglobulin

antibody form

-

antibody form

-

antibody form

affinity isolated antibody

shipped in

dry ice

shipped in

dry ice

shipped in

-

shipped in

wet ice

technique(s)

ELISA: suitable

technique(s)

ChIP: suitable, immunohistochemistry: suitable

technique(s)

-

technique(s)

direct ELISA: 1:30,000, dot blot: 1:30,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50, western blot: 1:30,000

concentration

4.59 mg/mL

concentration

-

concentration

-

concentration

-

manufacturer/tradename

Chemicon®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters