Skip to Content
Merck
All Photos(4)

Documents

MABN712

Sigma-Aldrich

Anti-CCL2 Antibody, clone 2D8

clone 2D8, from mouse

Synonym(s):

C-C motif chemokine 2, Monocyte chemoattractant protein 1, Monocyte chemotactic and activating factor, Monocyte chemotactic protein 1, Monocyte secretory protein JE, Small-inducible cytokine A2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D8, monoclonal

species reactivity

monkey, mouse, rat, human

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CCL2(6347)

General description

CCL2, Chemokine (C-C motif) ligand 2, or alternatively known as Monocyte chemotactic protein 1 (MCP-1), Monocyte chemotactic and activating factor (MCAF), Monocyte chemoattractant protein 1, Monocyte secretory protein JE, Small-inducible cytokine A2, HC11, and C-C motif chemokine 2 is encoded by the human gene named CCL2/MCP1/SCYA2. CCL2 is a chemokine produced by endothelial cells that attracts memory T cells, monocytes, dendritic cells, and basophils but not neutrophils or eosinophils to sites of inflammation produced by either tissue inquiry or infection. CCL2 also bolsters monocytes anti-tumor activity, and because it is chemotactic for monocytes, CCL2 has been implicated in inflammatory diseases where monocytic invasions have occurred such as in psoriasis, rheumatoid arthritis and atherosclerosis. CCL2 is a bifunctional molecule as well; the whole molecule is a basophil activator but removal of the N-terminal portion converts the molecule into an exclusive eosinophil chemoattractant. CCL2 can also be found expressed at the sites of tooth eruption and bone degradation and is expressed by both osteoclasts and osteoblasts. CCL2 is also expressed in neurons, astrocytes and microglia and its role in neuroinflammation is established. CCL2 expression in glial and neurons is increased in epilepsy, brain ischemia, Alzheimer’s disease, brain injury and experimental autoimmune encephalomyelitis. Finally, CCL2 plays a role in vascular complications associated with type II diabetes and in the production of amylin which contributes to insulin resistance in obesity, which is also associated with type II diabetes. EMD-Millipore’s Anti-CCL2 mouse monoclonal antibody has been tested in western blot against recombinant protein and cell lysates from A549, HeLa, Raw264.7, L1210, C6, and COS7 cell lines.

Immunogen

Purified recombinant fragment of human CCL2 expressed in E. Coli.

Application

Detect Monocyte Chemotactic Protein using this mouse monoclonal antibody, Anti-CCL2 Antibody, clone 2D8 validated for use in western blotting, Immunofluorescence, IHC & Flow Cytometry.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected CCL2 in HeLa cells.

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected CCL2 in human liver cancer tissue.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected CCL2 in A549 cells.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in A549, HeLa, Raw264.7, L1210, C6 and COS-7 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected CCL2 in A549, HeLa, Raw264.7, L1210, C6 and COS-7 cell lysates

Target description

~14 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Format: Purified
Purified mouse monoclonal in PBS with up to 0.1% sodium azide and 0.5% protein stabilizer.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A549, HeLa, Raw264.7, L1210, C6 and COS-7 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisha Luo et al.
Thoracic cancer, 13(6), 811-823 (2022-02-10)
Monocytes are involved in tumor growth and metastasis, but the distribution of monocyte phenotypes and their role in the development of malignant pleural effusion (MPE) remains unknown. A total of 94 MPE patients (76 diagnosed with adenocarcinoma lung cancer and
Milene Volpato et al.
Oncotarget, 7(19), 28139-28150 (2016-04-09)
The mechanism of the anti-colorectal cancer (CRC) activity of the omega-3 fatty acid eicosapentaenoic acid (EPA) is not understood. We tested the hypothesis that EPA reduces expression of chemokine C-C motif ligand 2 (CCL2), a pro-inflammatory chemokine with known roles
Xue Shi et al.
BMC neuroscience, 23(1), 20-20 (2022-04-01)
C-C motif chemokine ligand 2 (CCL2) is reported to be involved in the pathogenesis of various neurological and/or psychiatric diseases. Tissue or cellular expression of CCL2, in normal or pathological condition, may play an essential role in recruiting monocytes or
Aya Misawa et al.
Genes, 12(2) (2021-01-31)
Long noncoding RNAs (lncRNAs) are emerging as critical regulators of gene expression, which play fundamental roles in cancer development. In this study, we found that homeobox A11 antisense RNA (HOXA11-AS), a highly expressed lncRNA in cell lines derived from prostate
Anita Dittrich et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 45(2), 429-445 (2018-02-07)
Cardiovascular complications are common in astronauts returning from a prolonged spaceflight. These health problems might be driven by complex modulations of gene expression and protein synthesis in endothelial cells (ECs). Studies on the influence of microgravity on phenotype, growth pattern

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service