Skip to Content
Merck
All Photos(1)

Documents

MAB3307

Sigma-Aldrich

Anti-MMP-1 Antibody, a.a. 332-350 hMMP1, clone 41-1E5

clone 41-1E5, Chemicon®, from mouse

Synonym(s):

Fibroblast Collagenase, Interstitial Collagenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

41-1E5, monoclonal

species reactivity

rabbit, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MMP1(4312)

Specificity

Specifically reacts with precursor and active forms of human MMP-1.

Immunogen

Epitope: a.a. 332-350 hMMP1
Syntheic peptide aa332-350 of hMMP-1

Application

Anti-MMP-1 Antibody, a.a. 332-350 hMMP1, clone 41-1E5 detects level of MMP-1 & has been published & validated for use in ELISA, IH(P) & WB.
Immunoblotting:

Immunohistochemistry on PLP fixed, paraffin-embedded tissues 5-20μg/ml. PLP fixation required for best results. Antibody also reacts with frozen tissues. Reactivity on tradional formalin fixed tissue is poor. Microwave antigen retrieval is recommended

ElA

Target description

52 kDa

Linkage

Replaces: 04-1122

Physical form

Format: Purified
Liquid in 0.1 M Na-Phosphate buffer, pH 7.0 containing 2% protease free bovine Serum albumin.

Analysis Note

Control
Cervical carcinoma

SKBr-3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoru Kondo et al.
International journal of cancer, 115(3), 368-376 (2005-02-03)
The Epstein-Barr Virus (EBV) latent membrane protein 1 (LMP1) has a significant role in several malignancies, including nasopharyngeal carcinoma (NPC). LMP1 is the principal oncoprotein, and we have shown that it also induces a set of factors that mediates invasion
Carine Strup-Perrot et al.
World journal of gastroenterology, 11(40), 6312-6321 (2006-01-19)
To investigate their expression and activity in the rat ileum after exposure to ionizing radiation along with that of the cellular effectors and molecular mediators involved in the regulation of MMPs. Rats were exposed to a single 10-Gy dose of
Angela Ghatnekar et al.
The American journal of pathology, 182(6), 2391-2406 (2013-04-16)
Pulmonary arterial hypertension (PAH) is a chronic and progressive disease characterized by pulmonary vasculopathy with elevation of pulmonary artery pressure, often culminating in right ventricular failure. GATA-6, a member of the GATA family of zinc-finger transcription factors, is highly expressed
Jangyoun Choi et al.
International wound journal, 18(5), 647-656 (2021-03-19)
A hypertrophic scar is a common dermal fibroproliferative lesion usually treated with topical silicone. Verapamil, a type of calcium channel blocker, is considered a candidate drug for the treatment of hypertrophic scars. Here, we report that the addition of verapamil
Marc Irqsusi et al.
Journal of cardiac surgery, 37(6), 1613-1622 (2022-03-29)
The pathogenesis of mitral valve insufficiency is not yet fully understood. Several studies stressed the role of matrix metalloproteinases (MMPs) in the emergence of valvular pathologies. The primary objective of the present study is to analyze the role of selected MMPs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service