Skip to Content
Merck
All Photos(1)

Key Documents

CC327-1

Sigma-Aldrich

PhotoHA-LAP, Methacrylated Hyaluronic Acid Hydrogel Kit

Photocrosslinked hyaluronic acid hydrogel (CC327 Kit Component 1)

Synonym(s):

Hyaluronic Acid Scaffold, Methacrylated Hydrogel Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

form

lyophilized powder

application(s)

cell analysis

General description

3D cell culture, including bioprinting, allows for the creation of more physiological cell models by allowing cells to simultaneously interact with integrins on all cell surfaces, resulting in the activation of specific signaling pathways not activated in traditional 2D cell culture methods. Hydrogels are water swollen polymers that allow for the culture of cells in 3-dimensions and can have profound effects on cellular development, differentiation, migration, and function. New areas of tissue engineering such as 3D bioprinting, have utilized UV photocrosslinked methacrylated hydrogel biomaterials (PEGMA, GelMA, HAMA and ColMA etc.) to encapsulate cells to make printable bioinks. Hyaluronic acid is the most abundant glycosaminoglycan in the body being an important component of several tissues throughout the body. While it is abundant in extracellular matrices, hyaluronan also contributes to tissue hydrodynamics, movement and proliferation of cells, and participates in a number of cell surface receptor interactions. Hyaluronic acid is a polymer of disaccharides, themselves composed of D-glucuronic acid and Nacetyl- D-glucosamine, linked via alternating β-(1→4) and β-(1→3) glycosidic bonds. The PhotoHA-LAP, Methacrylated Hyaluronic Acid Hydrogel Kit (CC327: CC327-1, CC327-2) is based upon purified hyaluronic acid methacrylate (HAMA), which when photocrosslinked provides a native-like 3D environment for cells. In addition to hyaluronic acid methacrylate, the kit includes the photoinitiator lithium phenyl-2,4,6-trimethylbenzoylphosphinate (LAP) for users to easily fine tune their photocrosslinking experiments (i.e. altering hydrogel stiffness or gelling speeds).PhotoHA is a registered trademark of Advanced BioMatrix, Inc.

Application

Photocrosslinking is initiated using a 405 nm blue light source.

Packaging

1 Kit

Components

CC327-1 (Store at 2-8°C): LAP Photoinitiator, 1 X 100 mg (CS226445).
Note: The complete PhotoHA-LAP, Methacrylated Hyaluronic Acid Hydrogel Kit (CC327) consists of CC327-1 and CC327-2 sold seperately.

Quality

Appearance: Lyophilized PowderSterility (USP modified): No GrowthGrafting Efficiency: 50-70%pH: 6.0-8.0Osmolality: 200-400 mOsmo H20/kg in 1X PBSMolecular Weight: 100 – 150 kDaNMR: CharacteristicCell Compatibility: Characteristic

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Methacrylated hydrogels enable 3D cell culture scaffolds and bioprinting using light crosslinking.

Methacrylated hydrogels enable 3D cell culture scaffolds and bioprinting using light crosslinking.

Methacrylated hydrogels enable 3D cell culture scaffolds and bioprinting using light crosslinking.

Methacrylated hydrogels enable 3D cell culture scaffolds and bioprinting using light crosslinking.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service