8.56015
Fmoc-Phe-Wang resin (100-200 mesh)
Novabiochem®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352209
NACRES:
NA.22
Recommended Products
Quality Level
product line
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-30°C
General description
Pre-loaded resin for synthesis of peptide acids containing a C-terminal phenylalanine amino-acid residue by Fmoc SPPS.
This pre-loaded Wang resin is prepared using a synthesis-tested base resin, to ensure reproducibility and efficacy in SPPS. Loading of the Fmoc-amino acid is carried out under conditions to minimize enantiomerization and dipeptide formation, the values of which are provided on the COA. The lot number of the batch of Fmoc-amino acid used to prepare the resin is also provided on the COA, to provide transparency and traceability.
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
This pre-loaded Wang resin is prepared using a synthesis-tested base resin, to ensure reproducibility and efficacy in SPPS. Loading of the Fmoc-amino acid is carried out under conditions to minimize enantiomerization and dipeptide formation, the values of which are provided on the COA. The lot number of the batch of Fmoc-amino acid used to prepare the resin is also provided on the COA, to provide transparency and traceability.
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Linkage
Replaces: 04-12-2060
Analysis Note
Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (photometric determination of the Fmoc-chromophore liberated upon treatment with piperidine/DMF.): 0.20 - 1.00 mmol/g
Enantiomeric purity: ≥ 99.5 % (a/a)
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
Content Dipeptide (HPLC): ≤ 0.5 % (a/a)
Novabiochem Fmoc amino acid: lot number
The qualitiy of this resin has been confirmed by the successful synthesis of the 21mer peptide related to myelin oligodendrocyte glycoprotein (35-55), with an Fmoc-Val-Wang resin made from the same batch of base resin.
The polymer matrix is copoly (styrene - 1% DVB), 100 - 200 mesh.
Appearance of substance (visual): beads
Loading (photometric determination of the Fmoc-chromophore liberated upon treatment with piperidine/DMF.): 0.20 - 1.00 mmol/g
Enantiomeric purity: ≥ 99.5 % (a/a)
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
Content Dipeptide (HPLC): ≤ 0.5 % (a/a)
Novabiochem Fmoc amino acid: lot number
The qualitiy of this resin has been confirmed by the successful synthesis of the 21mer peptide related to myelin oligodendrocyte glycoprotein (35-55), with an Fmoc-Val-Wang resin made from the same batch of base resin.
The polymer matrix is copoly (styrene - 1% DVB), 100 - 200 mesh.
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service