Skip to Content
MilliporeSigma

Skip To

8.55014

NovaSyn®TG amino resin (130 µm)

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
$82.20
5 g
Check Cart for Availability
$326.00
25 g
Check Cart for Availability
$1,290.00

About This Item

NACRES:
NA.22
UNSPSC Code:
13111023

$82.20


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

NovaSyn® TG, Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

General description

NovaSyn TG amino resin is a composite of low cross-linked polystyrene and 3000-4000 M.W. polyethylene glycol [1,2,3], in which the PEG chains have been terminally functionalized with an amino group. The 130 μm beads have a narrow size distribution, high diffusion rates and excellent swelling across a wide range of solvents from toluene to water, making them ideally suited to the synthesis of serial and parallel libraries by organic solid phase synthesis. This resin can be easily converted into other supports suitable for use in SPOS by acylation with the appropriate carboxylic acid-containing linker. For example, Hutchins and Chapman [4] have coupled Riniker′s HMPB linker to this resin and used it to prepare a range of ureas derived from glutamic acid.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] W. Rapp, et al. in ′Innovation & Perspectives in Solid Phase Synthesis, 1st International Symposium′, R. Epton (Eds), SPCC UK Ltd., Birmingham, 1990, pp. 205.
[2] L. Zhang, et al. in ′Peptides 1990, Proc. 21st European Peptide Symposium′, E. Giralt & D. Andreu (Eds), ESCOM, Leiden, 1990, pp.196.
[3] E. Bayer (1991) Angew. Chem. Int. Ed. Engl., 30,113.
[4] S. M. Hutchins & K. T. Chapman (1994) Tetrahedron Lett., 35, 4055.

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.20 - 0.30 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
This resin is a composite of low-crossed linked polystyrene and 3000-4000 M.W. polyethylene glycol, which has been terminally amino functionalised. 130µm beads.

Other Notes

Replaces: 01-64-0094

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.550078.550638.55005
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

functional group

amine

functional group

amine

functional group

-

functional group

alcohol

product line

NovaSyn® TG, Novabiochem®

product line

NovaSyn® TG, Novabiochem®

product line

NovaSyn® TG, Novabiochem®

product line

NovaSyn® TG, Novabiochem®

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-30°C


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Hello, are these particles porous or nonporous? Thank you

    1 answer
    1. This particle is considered to be non-porous. The beads act as a separate gel phase rather than as a porous solid.

      Helpful?

Reviews

No rating value

Active Filters